Skip to Content

SML2397

Dooku1

≥98% (HPLC), eIF4E inhibitor, powder

Synonym(s):

2-((2,6-dichlorobenzyl)thio)-5-(1H-pyrrol-2-yl)-1,3,4-oxadiazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H9Cl2N3OS
CAS Number:
Molecular Weight:
326.20
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dooku1, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC(C=CC=C1Cl)=C1CSC2=NN=C(C3=CC=CN3)O2

InChI

1S/C13H9Cl2N3OS/c14-9-3-1-4-10(15)8(9)7-20-13-18-17-12(19-13)11-5-2-6-16-11/h1-6,16H,7H2

InChI key

MNPOBXLPCWFONX-UHFFFAOYSA-N

Biochem/physiol Actions

Dooku1, an analog of Yoda1, is a potent and selective antagonist of the mechanosensitive Piezo1 channel that inhibits endogenous Yoda1-activated channels. Dooku1 inhibits relaxation of relaxation of aorta caused by Yoda1.
potent and selective antagonist of the mechanosensitive Piezo1 channel that inhibits endogenous Yoda1-activated channels


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library