Skip to Content
Merck
All Photos(1)

Key Documents

SML2363

Sigma-Aldrich

AST487

≥98% (HPLC)

Synonym(s):

1-(4-((4-Ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-3-(4-(6-(methylamino)pyrimidin-4-yloxy)phenyl)urea, AST 487, AST-487, N-[4-[(4-Ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-N′-[4-[[6-(methylamino)-4-pyrimidinyl]oxy]phenyl]urea, NVP-AST 487, NVP-AST-487, NVP-AST487

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30F3N7O2
CAS Number:
Molecular Weight:
529.56
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCN1CCN(CC2=C(C(F)(F)F)C=C(NC(NC3=CC=C(OC4=NC=NC(NC)=C4)C=C3)=O)C=C2)CC1

InChI

1S/C26H30F3N7O2/c1-3-35-10-12-36(13-11-35)16-18-4-5-20(14-22(18)26(27,28)29)34-25(37)33-19-6-8-21(9-7-19)38-24-15-23(30-2)31-17-32-24/h4-9,14-15,17H,3,10-13,16H2,1-2H3,(H,30,31,32)(H2,33,34,37)

InChI key

ODPGGGTTYSGTGO-UHFFFAOYSA-N

Biochem/physiol Actions

AST487 is an orally available type II tyrosine kinase inhibitor (TKI) that exhibits potent antiproliferation activity against FLT3, RET, PDGFR1, but not BRAF, oncogenic fusions/mutations-driven growth (IC50 = 1.8 nM/FLT3-ITD BaF3, 5.1 nM/FLT3 D835I BaF3, 9-27 nM/TEL-PDGFR1 & PTC3-RET BaF3, <1 nM/FLT3-ITD-expressing MV4-11 &amp; MOLM-13) via binding/stablizing target kinases in the inactive “DFG-out” conformation. AST487 effectively suppresses RET- (NIH3T3-RETC634W & human medullary thyroid cancer (MTC) TT cells) and FLT3-ITD-driven (MV4-11) tumor growth in mice in vivo (20-50 mg/kg/day p.o.).
AST487 is a N,N′-diphenyl urea, which inhibits growth and calcitonin gene expression in medullary thyroid cancer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellen Weisberg et al.
Genes & cancer, 1(10), 1021-1032 (2011-07-23)
Constitutively activated mutant FLT3 has emerged as a promising target for therapy for the subpopulation of acute myeloid leukemia (AML) patients who harbor it. The small molecule inhibitor, PKC412, targets mutant FLT3 and is currently in late-stage clinical trials. However
Kristine K Deibler et al.
ACS chemical biology, 12(5), 1245-1256 (2017-03-07)
MEK4 is an upstream kinase in MAPK signaling pathways where it phosphorylates p38 MAPK and JNK in response to mitogenic and cellular stress queues. MEK4 is overexpressed and induces metastasis in advanced prostate cancer lesions. However, the value of MEK4
Ellen Weisberg et al.
Blood, 112(13), 5161-5170 (2008-09-30)
An attractive target for therapeutic intervention is constitutively activated, mutant FLT3, which is expressed in a subpopulation of patients with acute myelocyic leukemia (AML) and is generally a poor prognostic indicator in patients under the age of 65 years. PKC412
Matti L Gild et al.
Endocrine-related cancer, 20(5), 659-667 (2013-07-06)
Inhibitors of RET, a tyrosine kinase receptor encoded by a gene that is frequently mutated in medullary thyroid cancer, have emerged as promising novel therapies for the disease. Rapalogs and other mammalian target of rapamycin (mTOR) inhibitors are effective agents
Yu-Lin Hsieh et al.
Experimental neurology, 300, 87-99 (2017-11-07)
Neurotrophic factors and their corresponding receptors play key roles in the maintenance of different phenotypic dorsal root ganglion (DRG) neurons, the axons of which degenerate in small fiber neuropathy, leading to various neuropathic manifestations. Mechanisms underlying positive and negative symptoms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service