Skip to Content
Merck
All Photos(1)

Key Documents

SML2356

Sigma-Aldrich

A939572

≥98% (HPLC)

Synonym(s):

4-(2-Chlorophenoxy)-N-(3-(methylcarbamoyl)phenyl)piperidine-1-carboxamide, 4-(2-Chlorophenoxy)-N-[3-[(methylamino)carbonyl]phenyl]-1-piperidinecarboxamide, A 939572, A939

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22ClN3O3
CAS Number:
Molecular Weight:
387.86
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C20H22ClN3O3/c1-22-19(25)14-5-4-6-15(13-14)23-20(26)24-11-9-16(10-12-24)27-18-8-3-2-7-17(18)21/h2-8,13,16H,9-12H2,1H3,(H,22,25)(H,23,26)

InChI key

DPYTYQFYDLYWHZ-UHFFFAOYSA-N

Biochem/physiol Actions

A939572 is an orally available piperidine-aryl urea-based small molecule that inhibits stearoyl-CoA desaturase 1 (SCD1) with high potency (IC50 = 37 nM/hSCD1 and <4 nM/mSCD1) with little hERG channel blockade activity (IC50 >100 μM). When administered in ob/ob mice, A939572 effectively reduces desaturation indices in vivo (18:0/18:1n9 ratio from 26.9 to 8.47 in liver; from 15.4 to 7.35 in plasma post 5-day 10 mg/kg bid p.o.). A939572 is a useful tool for probing SCD1-mediated physiological and pathological processes both in cultures (50 nM-10 μM) and in vivo (5-10 mg/kg i.p. or 10 mg/kg p.o. in mice).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyeonwoo Kim et al.
Cell reports, 13(3), 495-503 (2015-10-13)
Some cancer cells exhibit elevated levels of free fatty acids (FAs) as well as high levels of β-catenin, a transcriptional co-activator that promotes their growth. Here, we link these two phenomena by showing that unsaturated FAs inhibit degradation of β-catenin.
Mohamed A Lounis et al.
American journal of physiology. Endocrinology and metabolism, 313(6), E710-E720 (2017-08-31)
Stearoyl-CoA desaturase-1 (SCD1) is a key player in lipid metabolism. SCD1 catalyzes the synthesis of monounsaturated fatty acids (MUFA). MUFA are then incorporated into triacylglycerols and phospholipids. Previous studies have shown that Scd1 deficiency in mice induces metabolic changes in
Ling Chen et al.
Scientific reports, 6, 19665-19665 (2016-01-28)
Inhibition of stearoyl-CoA desaturase 1 (SCD1) has been found to effectively suppress tumor cell proliferation and induce apoptosis in numerous neoplastic lesions. However, mechanism underlying SCD1-mediated anti-tumor effect has maintained unclear. Herein, we reported endo-lipid messenger ceramides played a critical
Mohamed A Lounis et al.
Biochimica et biophysica acta, 1861(11), 1662-1670 (2016-08-02)
Stearoyl-CoA desaturase 1 (SCD1) is a delta-9 fatty acid desaturase that catalyzes the synthesis of mono-unsaturated fatty acids (MUFA). SCD1 is a critical control point regulating hepatic lipid synthesis and β-oxidation. Scd1 KO mice are resistant to the development of
Mark Kin Fai Ma et al.
Journal of hepatology, 67(5), 979-990 (2017-06-26)
We investigated the functional role and clinical significance of stearoyl-CoA desaturase-1 (SCD1) mediated endoplasmic reticulum (ER) stress in regulating liver tumor-initiating cells (T-ICs) and sorafenib resistance, with the aim of developing a novel therapeutic strategy against hepatocellular carcinomas (HCCs). We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service