Skip to Content
Merck

SML2292

CCG-100602

≥98% (HPLC)

Synonym(s):

1-[3,5-Bis(trifluoromethyl)benzoyl]-N-(4-chlorophenyl)-3-piperidinecarboxamide, 1-[3,5-Bis(trifluoromethyl)benzoyl]-N-(4-chlorophenyl)piperidine-3-carboxamide, CCG 100602, CCG100602

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H17ClF6N2O2
CAS Number:
Molecular Weight:
478.82
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1CCCC(C(NC2=CC=C(Cl)C=C2)=O)C1)C3=CC(C(F)(F)F)=CC(C(F)(F)F)=C3

InChI

1S/C21H17ClF6N2O2/c22-16-3-5-17(6-4-16)29-18(31)12-2-1-7-30(11-12)19(32)13-8-14(20(23,24)25)10-15(9-13)21(26,27)28/h3-6,8-10,12H,1-2,7,11H2,(H,29,31)

InChI key

MOQCFMZWVKQBAP-UHFFFAOYSA-N

Biochem/physiol Actions

CCG-100602 aids in preventing fibrosis around the temporomandibular joint (TMJ).
CCG-100602 is a CCG-1423 analog with significantly less cytotoxicity (0/14% WST-1 inhibition by 10/100 μM CCG-100602 vs. 44% by 10 μM CCG-1423) and similar efficacy against Rho/MKL1/SRF pathway-mediated transcription (by 78% with 100 μM CCG-100602 vs. 74% with 10 μM CCG-1423; IC50 = 9.8 μM/CCG-100602 vs. 1.5 μM/CCG-1423; by PC-3 SRE-luciferase assay), albeit at a reduced potency. CCG-100602 reduces the stiffening of spontaneously hypertensive rats (SHR) derived aortic VSMCs (1.12 μM) by inhibiting SRF/myocardin interaction and abrogates the increased aortic wall stiffness in SHR rats in vivo (7.5 mg/kg/d via s.c. osmotic pumps for 2 wks).
CCG-1423 analog with similar efficacy and much less cytotoxicity. Inhibits SRF/myocardin interaction and reduces aortic VSMCs stiffening in vitro and in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Mark B Lucera et al.
Retrovirology, 14(1), 4-4 (2017-01-25)
HIV-1 hijacks host cell machinery to ensure successful replication, including cytoskeletal components for intracellular trafficking, nucleoproteins for pre-integration complex import, and the ESCRT pathway for assembly and budding. It is widely appreciated that cellular post-translational modifications (PTMs) regulate protein activity
Konstantin Tsoyi et al.
American journal of respiratory cell and molecular biology, 58(2), 208-215 (2017-09-09)
Radiation-induced pulmonary fibrosis is a severe complication of patients treated with thoracic irradiation. We have previously shown that syndecan-2 reduces fibrosis by exerting alveolar epithelial cytoprotective effects. Here, we investigate whether syndecan-2 attenuates radiation-induced pulmonary fibrosis by inhibiting fibroblast activation.
Ning Zhou et al.
Cardiovascular research, 113(2), 171-182 (2016-12-23)
Increased aortic stiffness is a fundamental manifestation of hypertension. However, the molecular mechanisms involved remain largely unknown. We tested the hypothesis that abnormal intrinsic vascular smooth muscle cell (VSMC) mechanical properties in large arteries, but not in distal arteries, contribute



Global Trade Item Number

SKUGTIN
SML2292-5MG04061835350155
SML2292-25MG04061835350148