Skip to Content
Merck

SML1867

NVS-PAK1-1

≥98% (HPLC)

Synonym(s):

(S)-3-(2-Chloro-5-(2,2-difluoroethyl)-8-fluoro-5H-dibenzo[b,e][1,4]diazepin-11-ylamino)-N-isopropylpyrrolidine-1-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H25ClF3N5O
CAS Number:
Molecular Weight:
479.93
UNSPSC Code:
41106609
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)CN1C2=C(C=C(Cl)C=C2)C(N[C@H]3CCN(C(NC(C)C)=O)C3)=NC4=C1C=CC(F)=C4

InChI

1S/C23H25ClF3N5O/c1-13(2)28-23(33)31-8-7-16(11-31)29-22-17-9-14(24)3-5-19(17)32(12-21(26)27)20-6-4-15(25)10-18(20)30-22/h3-6,9-10,13,16,21H,7-8,11-12H2,1-2H3,(H,28,33)(H,29,30)/t16-/m0/s1

InChI key

OINGHOPGNMYCAB-INIZCTEOSA-N

Biochem/physiol Actions

NVS-PAK1-1 is a potent allosteric inhibitor of p21-activated kinase PAK1, a serine-threonine kinase that is upregulated in many cancers.
NVS-PAK1-1 is a potent allosteric inhibitor of p21-activated kinase PAK1, a serine-threonine kinase that is upregulated in many cancers. NVS-PAK1-1 has an IC50 value of 5 nM for dephoshorylated PAK1 and 6 nM for phosphorylated PAK1 in an in vitro dephosphorylation assay. NVS-PAK1-1 showed good selectivity against 442 other kinases including PAK2. NVS-PAK1-1 has generally good physicochemical properties, but was metabolized by rat liver microsomes, so may not be useful for in vivo work. For full characterization details, please visit the NVS-PAK1-1 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for protein targets, visit sigma.com/sgc

Features and Benefits

NVS-PAK1-1 is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC
This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library