Skip to Content
Merck

SML1767

Sparstolonin B

≥98% (HPLC)

Synonym(s):

4,10-Dihydroxy-3H-pyrano[3,4,5-kl]xanthen-3-one, Sparstolenin B, SsnB

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H8O5
CAS Number:
Molecular Weight:
268.22
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C=C1)=CC2=C1OC3=C4C(C(OC=C42)=O)=C(O)C=C3

InChI

1S/C15H8O5/c16-7-1-3-11-8(5-7)9-6-19-15(18)14-10(17)2-4-12(20-11)13(9)14/h1-6,16-17H

InChI key

DHSASSJDMAHICE-UHFFFAOYSA-N

Biochem/physiol Actions

Potent TLR2 and TLR4 antagonist that exhibits anti-inflammatory and anti-viral (anti-HIV) properties
Sparstolonin B (SsnB), an isocoumarin isolated from Sparganium stoloniferum and Scirpus yagara, is a potent TLR2 and TLR4 antagonist that exhibits anti-inflammatory and anti-viral (anti-HIV) properties. Sparstolonin B inhibits LPS induced inflammation in 3T3- L1 adipocytes, and reduces the body weight of rats fed with high fat diet. Sparstolonin B mitigates inflammation in nonalcoholic steatohepatitis (NASH) in mice model.
Sparstolonin B (SsnB) is a polyphenol. It has a structural features similar to xanthone and isocoumarin. SsnB exerts anti-inflammatory properties on mouse and human macrophages. SsnB can significantly reduce hypoxia–reoxygenation-induced inflammation of cardiomyocytes by blocking extracellular signal-regulated protein kinase (ERK1/2) and c-jun N-terminal kinases (JNK) signaling pathways. Thus, SsnB acts as a potent therapeutic for the treatment of myocardial ischemia–reperfusion (MIR) injury.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library