Skip to Content
Merck

SML1745

Raptinal

≥95% (HPLC), mitochondrial pathway-mediated intrinsic apoptosis activator, powder

Synonym(s):

9H,9′H-9,9′-Bifluorene-9,9′-dicarbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H18O2
CAS Number:
Molecular Weight:
386.44
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Raptinal, ≥95% (HPLC)

Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=CC(C1=C2C=CC=C1)(C3(C=O)C4=C(C5=C3C=CC=C5)C=CC=C4)C6=C2C=CC=C6

InChI

1S/C28H18O2/c29-17-27(23-13-5-1-9-19(23)20-10-2-6-14-24(20)27)28(18-30)25-15-7-3-11-21(25)22-12-4-8-16-26(22)28/h1-18H

InChI key

GLANOOJJBKXTMI-UHFFFAOYSA-N

Application

Raptinal showed complete cytochrome c release within 30 minutes and caspase-3 activation within one hour of exposure (10 μM). Despite its general cytotoxicity in both cancer and non-cancer cell cultures (Cell death induction EC50 = 0.6-3.4 μM in 24 hr), no hematologic toxicity was observed 7 days following single 60 mg/kg i.v. dosing in mice. Studies show that daily i.p. injection (20 mg/kg) is efficacious in suppressing murine B16-F10 melanoma and 4T1 breast cancer expansion in vivo (by ~60% and 50% in 6 days, respectively).

Biochem/physiol Actions

A rapid activator of mitochondrial pathway-mediated intrinsic apoptosis
Raptinal is a cell-permeable bifluorene-dicarbaldehyde compound that acts as a rapid activator of mitochondrial pathway-mediated intrinsic apoptosis.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library