Skip to Content
Merck

Skip To

SML1665

Nocodazole

≥98% (HPLC), DMSO solution, microtubule inhibitor

Synonym(s):

Methyl N-[6-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate, Nocidazole, Nocodazolum, Oncodazole, R 17934

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H11N3O3S
CAS Number:
Molecular Weight:
301.32
UNSPSC Code:
12352200
NACRES:
NA.77
Form:
DMSO solution
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nocodazole Ready Made Solution, 5 mg/mL, DMSO solution

biological source

synthetic

Quality Level

form

DMSO solution

concentration

5 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[s]1c(ccc1)C(=O)c2cc3nc([nH]c3cc2)NC(=O)OC

InChI

1S/C14H11N3O3S/c1-20-14(19)17-13-15-9-5-4-8(7-10(9)16-13)12(18)11-3-2-6-21-11/h2-7H,1H3,(H2,15,16,17,19)

InChI key

KYRVNWMVYQXFEU-UHFFFAOYSA-N

Biochem/physiol Actions

Nocodazole is an anticancer drug that has been shown to interfere with the structure and function of microtubules in interphase and mitotic cells.
Nocodazole is an anticancer drug that has been shown to interfere with the structure and function of microtubules in interphase and mitotic cells. Malignant cells may be more susceptible to the antimicrotubular effect of nocodazole than nonmalignant cells. Mammalian cells cultured in vitro can be treated with 0.04-10 μg/mL doses for cell synchronization experiments. Prolonged arrest of cells in mitosis due to nocodazole treatment typically results in cell death by apoptosis. Higher concentrations could not be used because of insolubility. High specificity of action may explain low toxicity to bone marrow cells and lack of neurotoxicity. Nocodazole is thought to bind directly to tubulin causing conformational changes resulting in increased exposure of some sulfhydryl and possibly tyrosine residues. Nocodazole′s apparent synergism with cytosine arabinofuranoside has been demonstrated on L1210 leukemic cells.

Preparation Note

Nocodazole forms precipitates in frozen solution, therefore sonication should be applied after thaw to homogenize the solution. It is recommended to sonicate for 1-3 minutes post thawing to remove precipitates that might appear due to freezing.
For cell culture work, dilute Nocodazole ready-made solution (5 mg/mL DMSO) in the culture media, diluted solution is stable for at least 7 days in biological. Mammalian cells cultured in vitro can be treated with 0.04-10 μg/mL doses.
The product is stable for 2 years at -20C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M1404487928SML0077
form

DMSO solution

form

powder

form

solid

form

powder

Quality Level

200

Quality Level

300

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

10-30°C

storage temp.

2-8°C

shipped in

dry ice

shipped in

-

shipped in

ambient

shipped in

-

concentration

5 mg/mL

concentration

-

concentration

-

concentration

-

biological source

synthetic

biological source

-

biological source

-

biological source

-


Still not finding the right product?

Explore all of our products under Nocodazole Ready Made Solution


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Anne Meldgaard Hansen et al.
Science advances, 8(11), eabf8627-eabf8627 (2022-03-19)
Activation of interferon genes constitutes an important anticancer pathway able to restrict proliferation of cancer cells. Here, we demonstrate that the H3K9me3 histone methyltransferase (HMT) suppressor of variegation 3-9 homolog 1 (SUV39H1) is required for the proliferation of acute myeloid
Francesco Cambuli et al.
EMBO reports, 23(5), e54049-e54049 (2022-03-08)
The healthy prostate is a relatively quiescent tissue. Yet, prostate epithelium overgrowth is a common condition during aging, associated with urinary dysfunction and tumorigenesis. For over thirty years, TGF-β ligands have been known to induce cytostasis in a variety of
Anne Monette et al.
Cell reports, 40(8), 111251-111251 (2022-08-25)
Membraneless biomolecular condensates (BMCs) contribute to the replication of a growing number of viruses but remain to be functionally characterized. Previously, we demonstrated that pan-retroviral nucleocapsid (NC) proteins phase separated into condensates regulating virus assembly. Here we discover that intrinsically



Global Trade Item Number

SKUGTIN
SML1665-1ML04061838093035

Questions

Reviews

No rating value

Active Filters