Skip to Content
Merck
All Photos(1)

Key Documents

SML1476

Sigma-Aldrich

Thiocolchicoside

≥98% (HPLC)

Synonym(s):

(S)-N-[3-(β-D-glucopyranosyloxy)-1,2-dimethoxy-10(methylthio)-9-oxo-5,6,7,9-tetrahydrobenzo [a] heptalen-7-yl] acetamide, 2-Demethoxy-2-glucosidoxythiocolchicine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H33NO10S
CAS Number:
Molecular Weight:
563.62
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -510 to -610°, c = 0.5 in H2O

color

light yellow to dark yellow

solubility

H2O: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](OC2=CC(CC[C@H](NC(C)=O)C(C3=CC=C4SC)=CC4=O)=C3C(OC)=C2OC)[C@@H]1O

InChI

1S/C27H33NO10S/c1-12(30)28-16-7-5-13-9-18(37-27-24(34)23(33)22(32)19(11-29)38-27)25(35-2)26(36-3)21(13)14-6-8-20(39-4)17(31)10-15(14)16/h6,8-10,16,19,22-24,27,29,32-34H,5,7,11H2,1-4H3,(H,28,30)/t16-,19+,22+,23-,24+,27+/m0/s1

InChI key

LEQAKWQJCITZNK-AXHKHJLKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Thiocolchicoside acts as a muscle relaxant. In experimental animals, thiocolchicoside results in malfunctions (teratogenic) and impairs chromosomes. It is advised to be used as an analgesic for patients with muscle pain, starting with paracetamol. Thiocolchicoside can be used in irrigation solution at the time of endoscopic treatment of ureteral calculus to reduce the movement of stones.
Thiocolchicoside is a potent competitive γ-aminobutyric acid type A (GABAA) receptor antagonist and glycine receptor agonist with muscle relaxant, anti-inflammatory, and analgesic activity. Thiocolchicoside has also been found to suppress nuclear factor kappa B ligand (RANKL) activation of NF-KB resulting in suppression of osteoclastogenesis.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2 - STOT RE 1

Target Organs

Gastro-intestinal system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiocolchicoside: review of adverse effects.
Prescrire International, 25(168), 3-41 (2016)
Impact of using thiocolchicoside during endoscopic ureteral calculi removal: A preliminary study.
Akin Y, et al.
Minimally Invasive Therapy & Allied Technologies : MITAT : Official Journal of the Society for Minimally Invasive Therapy, 25(1), 29-34 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service