Skip to Content
Merck
All Photos(1)

Key Documents

SML1438

Sigma-Aldrich

LY2090314

≥98% (HPLC)

Synonym(s):

3-[9-Fluoro-1,2,3,4-tetrahydro-2-(1-piperidinylcarbonyl)pyrrolo[3,2,1-jk][1,4]benzodiazepin-7-yl]-4-imidazo[1,2-a]pyridin-3-yl-1h-pyrrole-2,5-dione, 3-[9-Fluoro-2-(piperidin-1-ylcarbonyl)-1,2,3,4-tetrahydro[1,4]diazepino[6,7,1-hi]indol-7-yl]-4-imidazo[1,2-a]pyridin-3-yl-1H-pyrrole-2,5-dione, LY-2090314

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H25FN6O3
CAS Number:
Molecular Weight:
512.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1CCCCC1)N(C2)CCN3C4=C2C=C(F)C=C4C(C5=C(C6=CN=C7N6C=CC=C7)C(NC5=O)=O)=C3

InChI

1S/C28H25FN6O3/c29-18-12-17-15-34(28(38)32-7-3-1-4-8-32)11-10-33-16-20(19(13-18)25(17)33)23-24(27(37)31-26(23)36)21-14-30-22-6-2-5-9-35(21)22/h2,5-6,9,12-14,16H,1,3-4,7-8,10-11,15H2,(H,31,36,37)

InChI key

HRJWTAWVFDCTGO-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

LY2090314 can decrease the development of human MYCN amplified and non-amplified neuroblastoma (NB) cell lines in vitro. It has the ability to prevent the multiplication, colony formation and cell confluency of neuroblastoma.
LY2090314 is a potent and selective ATP-competitive inhibitor of Glycogen synthase kinase-3 (GSK-3) currently in clinical trials for cancer therapy. LY2090314 has IC50 values of 1.5 nM and 0.9 nM for GSK-3α and GSK-3β, respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antiproliferative and apoptotic effect of LY2090314, a GSK-3 inhibitor, in neuroblastoma in vitro
Kunnimalaiyaan S, et al.
BMC Cancer, 18(1), 560-560 (2018)
Mengfei Yu et al.
Cell, 184(1), 243-256 (2021-01-09)
Craniosynostosis results from premature fusion of the cranial suture(s), which contain mesenchymal stem cells (MSCs) that are crucial for calvarial expansion in coordination with brain growth. Infants with craniosynostosis have skull dysmorphology, increased intracranial pressure, and complications such as neurocognitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service