Skip to Content
Merck
All Photos(1)

Documents

SML1318

Sigma-Aldrich

Ingenol-3-angelate

≥95% (HPLC)

Synonym(s):

(1aR,2S,5R,5aS,6S,8aS,9R,10aR)-5,5a-Dihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-11-oxo-1a,2,5,5a,6,9,10,10a-octahydro-1H-2,8a-methanocyclopenta[a]cyclpropa[e][10]annulen-6-yl (2Z)-2-methylbut-2-enoate, 3-Angeloylingenol, 3-Ingenyl angelate, I3A, Ingenol mebutate, PEP005

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34O6
CAS Number:
Molecular Weight:
430.53
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

semisynthetic

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1[C@@]2(C=C(C)[C@@H]3OC(/C(C)=C\C)=O)[C@]3(O)[C@H](O)C(CO)=C[C@@]1([H])[C@@](C4(C)C)([H])[C@@]4([H])C[C@H]2C

InChI

1S/C25H34O6/c1-7-12(2)22(29)31-21-13(3)10-24-14(4)8-17-18(23(17,5)6)16(20(24)28)9-15(11-26)19(27)25(21,24)30/h7,9-10,14,16-19,21,26-27,30H,8,11H2,1-6H3/b12-7-/t14-,16+,17-,18+,19-,21+,24+,25+/m1/s1

InChI key

VDJHFHXMUKFKET-WDUFCVPESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ingenol 3-Angelate is a diterpenoid ester isolated from Euphorbia peplus that induces rapid cell death with keratinocytes and some cancer cell lines. Ingenol 3-Angelate is a protein kinase C activator that displays potent antileukemic activity. Ingenol 3-Angelate (ingenol mebutate) is used for treatment of actinic keratosis.
Ingenol-3-angelate, is a phorbol ester-like compound or a diacylglycerol analog that is used in the treatment of several disorders like skin cancer. It is also known as PEP005. Ingenol-3-angelate stimulates the initiation of the enzyme by providing PKCs (protein kinase C) to cellular membranes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yunci Ma et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 113, 108753-108753 (2019-03-12)
The objective of this study was to investigate the effects of ferulic acid (FA) on nonalcoholic fatty liver disease (NAFLD) and gut microbiota, and its regulation mechanism in ApoE-/- mice fed on a high-fat diet (HFD). Liver morphology, blood lipids
Shen Lin et al.
PloS one, 12(5), e0177496-e0177496 (2017-05-16)
The role of posttranslational modifications in axonal injury and regeneration has been widely studied but there has been little consensus over the mechanism by which each modification affects adult axonal growth. Acetylation is known to play an important role in
Xiaohua Song et al.
PloS one, 8(8), e72331-e72331 (2013-08-31)
Ingenol-3-angelate (I3A) is a non-tumor promoting phorbol ester-like compound identified in the sap of Euphoria peplus. Similar to tumor promoting phorbol esters, I3A is a diacylglycerol (DAG) analogue that binds with high affinity to the C1 domains of PKCs, recruits
Parama Paul et al.
PloS one, 14(5), e0216220-e0216220 (2019-05-07)
Autosomal dominant polycystic kidney disease (ADPKD) is caused mostly by mutations in polycystin-1 or polycystin-2. Fluid flow leads to polycystin-dependent calcium influx and nuclear export of histone deacetylase 5 (HDAC5), which facilitates the maintenance of renal epithelial architecture by de-repression
Mian Yu et al.
Journal of controlled release : official journal of the Controlled Release Society, 304, 242-258 (2019-05-10)
Cancer-Immunotherapy was the most exciting topic. However, either insensitivity due to singleness of therapeutic target or immune evasion leads to the failure of the treatment. Ingenol-3-mebutate (I3A) can inhibit cancer through synergy between immunotherapy and chemotherapy, however, the speculation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service