Skip to Content
Merck
All Photos(1)

Key Documents

SML1314

Sigma-Aldrich

SAG dihydrochloride

≥98% (HPLC), lyophilized solid, smoothened (Smo) agonist

Synonym(s):

3-Chloro-N-[trans-4-(methylamino)cyclohexyl]-N-[[3-(4-pyridinyl)phenyl]methyl]-benzo[b]thiophene-2-carboxamide dihydrochloride, Smoothened Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28ClN3OS · 2HCl
CAS Number:
Molecular Weight:
562.98
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

product name

SAG dihydrochloride, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(C1=C(Cl)C2=C(C=CC=C2)S1)N([C@@]3([H])CC[C@@](NC)([H])CC3)CC4=CC(C5=CC=NC=C5)=CC=C4

InChI

1S/C28H28ClN3OS/c1-30-22-9-11-23(12-10-22)32(28(33)27-26(29)24-7-2-3-8-25(24)34-27)18-19-5-4-6-21(17-19)20-13-15-31-16-14-20/h2-8,13-17,22-23,30H,9-12,18H2,1H3/t22-,23-

InChI key

VFSUUTYAEQOIMW-YHBQERECSA-N

Application

Smoothened agonist (SAG) dihydrochloride has been used as a sonic hedgehog (SHH) pathway agonist:
  • to study its effects on mouse hippocampal cells
  • to determine its effects on the mitochondrial network in zebrafish embryo
  • to study its effects on murine embryonic fibroblasts

Biochem/physiol Actions

SAG is a potent and cell-permeable smoothened (Smo) agonist that activates Hedgehog pathway and counteracts Cyclopamine inhibition of Smo.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela Prochazkova et al.
Development (Cambridge, England), 144(12), 2212-2221 (2017-05-17)
The patterning of repeated structures is a major theme in developmental biology, and the inter-relationship between spacing and size of such structures is an unresolved issue. Fungiform papillae are repeated epithelial structures that house taste buds on the anterior tongue.
Alp Ozgun et al.
Materials today. Bio, 25, 100992-100992 (2024-02-19)
Modern in vitro technologies for preclinical research, including organ-on-a-chip, organoids- and assembloid-based systems, have rapidly emerged as pivotal tools for elucidating disease mechanisms and assessing the efficacy of putative therapeutics. In this context, advanced in vitro models of Parkinson's Disease
Lina Zhu et al.
PeerJ, 10, e13381-e13381 (2022-05-10)
Smoothened (SMO) protein is a member of the G protein-coupled receptor (GPCR) family that is involved in the Hedgehog (Hh) signaling pathway. It is a putative target for treating various cancers, including medulloblastoma and basal cell carcinoma (BCC). Characterizing membrane
Shirin Farheen et al.
Journal of biochemical and molecular toxicology, 37(2), e23241-e23241 (2022-10-08)
Glioma is a major brain tumor, and the associated mortality rate is very high. Contemporary therapies provide a chance of survival for 9-12 months. Therefore, a novel approach is essential to improve the survival rate. Sonic hedgehog (Shh) cell signaling
Maike Marczenke et al.
Development (Cambridge, England), 148(21) (2021-10-27)
Growth arrest-specific 1 (GAS1) acts as a co-receptor to patched 1, promoting sonic hedgehog (SHH) signaling in the developing nervous system. GAS1 mutations in humans and animal models result in forebrain and craniofacial malformations, defects ascribed to a function for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service