Skip to Content
Merck
All Photos(1)

Key Documents

SML1269

Sigma-Aldrich

Cepharanthine

≥95% (HPLC)

Synonym(s):

(+)-Cepharanthine, 6′,12′-Dimethoxy-2,2′-dimethyl-6,7-(methylenebis(oxy))oxyacanthan, O-Methylcepharanoline, CEP, Cepharanthin, Cepharantin, NSC 623442, [14aS-(14aR*,26aS*)]-2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl- H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2′,3′:17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H38N2O6
CAS Number:
Molecular Weight:
606.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

(isolated from the tuber of Stephania Cepharantha Hayata (plant).)

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

CN1CCC2=CC3=C(OCO3)C(O4)=C2[C@@H]1CC5=CC=C(OC6=C(OC)C=CC(C[C@@H]7C8=CC4=C(OC)C=C8CCN7C)=C6)C=C5

InChI

1S/C37H38N2O6/c1-38-13-11-24-18-31(41-4)33-20-27(24)28(38)16-23-7-10-30(40-3)32(17-23)44-26-8-5-22(6-9-26)15-29-35-25(12-14-39(29)2)19-34-36(37(35)45-33)43-21-42-34/h5-10,17-20,28-29H,11-16,21H2,1-4H3/t28-,29+/m1/s1

InChI key

YVPXVXANRNDGTA-WDYNHAJCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cepharanthine is an anti-inflammatory, antiallergic, immunomo- modulatory and antineoplastic alkaloid isolated from Stephania cepharantha Hayata. Cepharanthine is an inhibitor of efflux transporter ABCC10. Also, Cepharanthine binds to central portion of Hsp90 with Kd 15 nM. Recent studies suggest that Cepharanthine inhibits the entry of HIV-1 by reducing plasma membrane fluidity.
Cepharanthine is known to be the only biscoclaurine alkaloid containing a methylenedioxy group. It promotes leukocytosis, as a result, it might activate the reticular endothelial system, hematopoietic tissue and bone marrow hyperplasia. It is known to inhibit Ehrlich tumor proliferation. Cepharanthine can increase the sensitivity and efficacy of antitumor drugs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kouki Matsuda et al.
Bioorganic & medicinal chemistry letters, 24(9), 2115-2117 (2014-04-08)
The anti-HIV-1 activity of cepharanthine (CEP), a natural product derived from Stephania cepharantha Hayata, was evaluated. CEP stabilized plasma membrane fluidity and inhibited HIV-1 envelope-dependent cell-to-cell fusion of HIV-1-infected cells as well as cell-free infection. It is suggested that CEP
The Alkaloids, 223-223 (2014)
Rishil J Kathawala et al.
Chinese journal of cancer, 33(5), 223-230 (2013-10-10)
ABCC10, also known as multidrug-resistant protein 7 (MRP7), is the tenth member of the C subfamily of the ATP-binding cassette (ABC) superfamily. ABCC10 mediates multidrug resistance (MDR) in cancer cells by preventing the intracellular accumulation of certain antitumor drugs. The
Natural Small Molecule Drugs from Plants, 499-499 (2018)
Jun Haginaka et al.
Analytical biochemistry, 434(1), 202-206 (2012-12-12)
Heat shock protein 90α (Hsp90α) immobilized on aminopropyl silica gels was prepared via the N- or C-terminal, which was termed Hsp90α-NT or Hsp90α-CT, respectively. Binding interactions of biscoclaurine alkaloids (cepharanthine (CEP), berbamine (BBM), isotetrandrine (ITD), and cycleanine (CCN)) with Hsp90α

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service