Skip to Content
Merck
All Photos(1)

Documents

SML1175

Sigma-Aldrich

Romidepsin

≥98% (HPLC), powder, histone deacetylase inhibitor

Synonym(s):

Cyclo[(2Z)-2-amino-2-butenoyl-L-valyl-(3S,4E)-3-hydroxy-7-mercapto-4-heptenoyl-D-valyl-D-cysteinyl] cyclic (35)-disulfide, Depsipeptide, FK228, FR901228, L-Valine, N-((3S,4E)-3-hydroxy-7-mercapto-1-oxo-4-heptenyl)-D-valyl-D-cysteinyl-(2Z)-2-amino-2-butenoxyl-, (4-1)-lactone, cyclic (1-2)-disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36N4O6S2
CAS Number:
Molecular Weight:
540.70
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Romidepsin, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N/C(C(N[C@@H](C(C)C)C(O[C@@]1([H])CC2=O)=O)=O)=C\C)[C@@H](CSSCC/C=C/1)NC([C@H](N2)C(C)C)=O

InChI

1S/C24H36N4O6S2/c1-6-16-21(30)28-20(14(4)5)24(33)34-15-9-7-8-10-35-36-12-17(22(31)25-16)26-23(32)19(13(2)3)27-18(29)11-15/h6-7,9,13-15,17,19-20H,8,10-12H2,1-5H3,(H,25,31)(H,26,32)(H,27,29)(H,28,30)/b9-7+,16-6-/t15-,17-,19-,20+/m1/s1

InChI key

OHRURASPPZQGQM-GCCNXGTGSA-N

Biochem/physiol Actions

Romidepsin is a very potent natural prodrug inhibitor of HDAC1 and HDAC2 that is converted to active form by glutathione. Romidepsin has IC50 values of 36 nM and 47 nM for HDAC1 and HDAC2, respectively. Romidepsin kills lymphoma cell lines overexpressing Bcl-2 and Bcl-XL, and has been approved for the treatment for cutaneous T-cell lymphoma (CTCL) and peripheral T-cell lymphoma, and a variety of other cancers.
Romidepsin promotes apoptosis, while hindering tumor growth in glioma models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Otte et al.
International journal of oncology, 49(6), 2453-2463 (2016-10-18)
Chemotherapeutic drug testing of SCCOHT-1 and BIN-67 tumor cells revealed synergistic growth-inhibition of >95% in vitro with a combination of foretinib and FK228. Application of this drug combination in vivo in NODscid mice-induced SCCOHT-1GFP tumors was associated with ~6-fold reduction in
A phase I/II trial of the histone deacetylase inhibitor romidepsin for adults with recurrent malignant glioma: North American Brain Tumor Consortium Study 03-03.
Iwamoto F M, et al.
Neuro-Oncology, 13(5), 509-516 (2011)
Sara Moron-Lopez et al.
AIDS (London, England), 33(3), 425-431 (2018-12-12)
Reversing HIV-1 latency has been suggested as a strategy to eradicate HIV-1. We investigated the effect of romidepsin on the HIV transcription profile in participants from the REDUC part B clinical trial. Seventeen participants on suppressive antiretroviral therapy were vaccinated
Minsun Jung et al.
The oncologist, 24(8), e740-e748 (2019-01-31)
NUT carcinoma is a rare aggressive disease caused by BRD4/3-NUT fusion, and C-MYC upregulation plays a key role in the pathogenesis. Here, we report on the clinicopathological characteristics of Korean patients with NUT carcinoma and the in vitro efficacy of
Cai-Ping Chen et al.
Acta pharmaceutica Sinica. B, 9(5), 937-951 (2019-10-28)
Artemisinin and its derivatives (ARTs) were reported to display heme-dependent antitumor activity. On the other hand, histone deacetylase inhibitors (HDACi) were known to be able to promote heme synthesis in erythroid cells. Nevertheless, the effect of HDACi on heme homeostasis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service