Skip to Content
Merck
All Photos(1)

Key Documents

SML1092

Sigma-Aldrich

Levobupivacaine hydrochloride

≥98% (HPLC)

Synonym(s):

(S)-1-Butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide hydrochloride, (S)-bupivacaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O · HCl
CAS Number:
Molecular Weight:
324.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -10 to -14°, c = 1.0 in H2O

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(NC([C@H]2N(CCCC)CCCC2)=O)C(C)=CC=C1.Cl

InChI

1S/C18H28N2O.ClH/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H/t16-;/m0./s1

InChI key

SIEYLFHKZGLBNX-NTISSMGPSA-N

Gene Information

human ... SCN4A(6329)

General description

Levobupivacaine belongs to the N-alkyl substituted pipecoloxylidide family and comprises amino-amide group.

Application

Levobupivacaine hydrochloride has been used as an analyte in tandem mass spectrometry. It may be used to test its inhibitory effect on phosphorylation of extracellular signal-regulated kinase (ERK) mediated by capsaicin It may also be used as a component of poly(D,L-lactide-co-glycolide) (PLGA) microparticles for testing its sustainable release by electrospraying technique

Biochem/physiol Actions

Levobupivacaine hydrochloride is a sodium channel blocker used as a long-acting local anaesthetic for epidural anesthesia. Levobupivacaine is the (S)-isomer of bupivacaine, with efficacy similar to that of bupivacaine with a reduced risk of cardiotoxicity.
Levobupivacaine hydrochloride is a sodium channel blocker.
Levobupivacaine mediates its action by blocking the action potential and sodium passage in neuronal membrane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Different effects of local anesthetics on extracellular signal-regulated kinase phosphorylation in rat dorsal horn neurons
Zhang L, et al.
European Journal of Pharmacology, 734(6), 132-136 (2014)
Salima Qudsi-Sinclair et al.
Pain practice : the official journal of World Institute of Pain, 17(5), 578-588 (2016-09-20)
Knee osteoarthritis is a disease that affects a third of the population over 65 years of age, and it is increasingly becoming a motive for consultation and a source of pain and disability. The gold standard surgical treatment is a total
Joanne Guay et al.
The Cochrane database of systematic reviews, 3, CD006715-CD006715 (2019-03-02)
General anaesthesia combined with epidural analgesia may have a beneficial effect on clinical outcomes. However, use of epidural analgesia for cardiac surgery is controversial due to a theoretical increased risk of epidural haematoma associated with systemic heparinization. This review was
Tianci Li et al.
Journal of anesthesia, 33(1), 17-25 (2018-11-15)
Retrospective studies indicate that the use of regional anaesthesia causes a reduction in cancer recurrence after oncological surgery, which could be due to anaesthetic's negating effect on immunosuppression related to the surgical stress response. Local anaesthetics may also exert direct
Jian-Ming Chen et al.
International journal of molecular sciences, 21(3) (2020-02-12)
In this study, we explored the release characteristics of analgesics, namely levobupivacaine, lidocaine, and acemetacin, from electrosprayed poly(D,L-lactide-co-glycolide) (PLGA) microparticles. The drug-loaded particles were prepared using electrospraying techniques and evaluated for their morphology, drug release kinetics, and pain relief activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service