Skip to Content
Merck

SML1079

BMOV

≥95% (elemental analysis)

Synonym(s):

(SP-5-31)-bis(3-hydroxy-2-methyl-4H-pyran-4-onato-O3,O4)oxo-vanadium, Bis(maltolato)oxovanadium(IV)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H10O7V
CAS Number:
Molecular Weight:
317.15
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥95% (elemental analysis)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (elemental analysis)

form

powder

color

, yellow to grey to brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[V+2]=O.O2[C-](C(=O)C(=O)C=C2)C.O1[C-](C(=O)C(=O)C=C1)C

InChI

1S/2C6H5O3.O.V/c2*1-4-6(8)5(7)2-3-9-4;;/h2*2-3H,1H3;;/q2*-1;;+2

InChI key

NGWOELASSYWDDT-UHFFFAOYSA-N

Biochem/physiol Actions

BMOV is a potent, orally available vanadium complex that acts as insulin mimetic. Apparently, BMOV anti-diabetic effect results from enhancement of the tyrosine-phosphorylation of insulin receptor by inhibition of the protein tyrosine phosphatase-1B (PTP-1B). In diabetic rats BMOV restores normoglycaemia without rising insulin levels. BMOV is a potent phosphatase inhibitor.
BMOV is a potent, orally available vanadium complex that acts as insulin mimetic; anti-diabetic; potent phosphatase inhibitor.


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library