Skip to Content
Merck

SML1045

Thiazovivin

≥98% (HPLC), powder, Rho Kinase inhibitor

Synonym(s):

N-Benzyl-2-(pyrimidin-4-ylamino)thiazole-4-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H13N5OS
CAS Number:
Molecular Weight:
311.36
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Thiazovivin, ≥98% (HPLC)

Quality Level

assay

≥98% (HPLC)

form

powder

color

, white to beige to brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1c(nc(c1)C(=O)NCc3ccccc3)Nc2ncncc2

InChI

1S/C15H13N5OS/c21-14(17-8-11-4-2-1-3-5-11)12-9-22-15(19-12)20-13-6-7-16-10-18-13/h1-7,9-10H,8H2,(H,17,21)(H,16,18,19,20)

InChI key

DOBKQCZBPPCLEG-UHFFFAOYSA-N

Application

Thiazovivin has been used in the generation of induced pluripotent stem cells (iPSCs) and induced neural stem cells (iNSCs) from human urine cells. It has also been used to study the the effect of pro-fibrotic inhibition on cardiac reprogramming.

Biochem/physiol Actions

Thiazovivin is a Rho Kinase (ROCK) inhibitor that promotes the transformation of fibroblasts into stem cells with a 200-fold efficiency over the classic method when used in combination with ALK5 inhibitor SB-431542 (S4317) and MEK inhibitor PD-0325901 (PZ0162). Thiazovivin stabilizes E-cadherin on the cell surface, necessary for human embryonic stem cell survival in culture. When human embryonic stem cells are cut out from the colony, this key protein is disrupted and then internalized within the cell. Without e-cadherin on the cell surface, cell signaling between the cells and their environment is disrupted and the cells quickly die.
Thiazovivin is a Rho Kinase (ROCK) inhibitor.
Thiazovivin is an inhibitor of Rho associated coiled-coil containing protein kinase (ROCK). In vitro studies prove that thiazovivin is efficient in stimulating better morphology, expression of ionic transporter and protein involved in cell adhesion.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Generation of Urine Cell-Derived Non-integrative Human iPSCs and iNSCs: A Step-by-Step Optimized Protocol.
Cheng L, et al.
Frontiers in Molecular Neuroscience, 10, 348-348 (2017)
The ROCK inhibitor, thiazovivin, inhibits human corneal endothelial-to-mesenchymal transition/epithelial-to-mesenchymal transition and increases ionic transporter expression.
Wu Q, et al.
International Journal of Molecular Medicine, 40(4), 1009-1018 (2017)
Daniel A Armendariz et al.
eLife, 12 (2023-04-25)
Enhancers orchestrate gene expression programs that drive multicellular development and lineage commitment. Thus, genetic variants at enhancers are thought to contribute to developmental diseases by altering cell fate commitment. However, while many variant-containing enhancers have been identified, studies to endogenously



Global Trade Item Number

SKUGTIN
SML1045-25MG04061832451428
SML1045-5MG04061832451435