Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C24H23ClO2
CAS Number:
Molecular Weight:
378.89
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 20 mg/mL, clear
storage temp.
2-8°C
SMILES string
ClCC\C(=C(\c3ccc(cc3)OCCO)/c2ccccc2)\c1ccccc1
InChI
1S/C24H23ClO2/c25-16-15-23(19-7-3-1-4-8-19)24(20-9-5-2-6-10-20)21-11-13-22(14-12-21)27-18-17-26/h1-14,26H,15-18H2/b24-23-
InChI key
LUMKNAVTFCDUIE-VHXPQNKSSA-N
Biochem/physiol Actions
Ospemifene is a selective estrogen receptor modulator (SERM), a metabolite of toremifene, with a unique estrogen agonist/antagonist tissue profile. Ospemifene was recently approved in the US for the treatment of dyspareunia associated with vulvar and vaginal atrophy in postmenopausal women, and is being investigated for the treatment and prevention of osteoporosis and breast cancer. Ospemifene binds ERα and ERβ with approximately equal affinities with IC50 values for estrogen receptor (ER) α and β of 0.8 μM and 1.7 μM, respectively. Ospemifene has estrogen-like effects on the vaginal epithelium and bone, antiproliferative effects in breast, and a neutral endometrial profile.
Ospemifene is a selective estrogen receptor modulator (SERM), a metabolite of toremifene.
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
