Skip to Content
Merck

SML0993

MK-677

≥98% (HPLC)

Synonym(s):

Crescendo, Ibutamoren mesylate, L-163191, MK-0677, MK-667, N-[1(R)-1,2-Dihydro-1-methanesulfonylspiro-3H-indole-3,4′-piperidin)-1′-yl]carbonyl-2-(phenylmethoxy)-ethyl-2-amino-2-methylpropanamide methanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H36N4O5S · CH3SO3H
CAS Number:
Molecular Weight:
624.77
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N1CC3(CCN(CC3)C(=O)[C@H](NC(=O)C([N+H3])(C)C)COCc4ccccc4)c2c1cccc2)C.[S](=O)(=O)([O-])C

InChI

1S/C27H36N4O5S.CH4O3S/c1-26(2,28)25(33)29-22(18-36-17-20-9-5-4-6-10-20)24(32)30-15-13-27(14-16-30)19-31(37(3,34)35)23-12-8-7-11-21(23)27;1-5(2,3)4/h4-12,22H,13-19,28H2,1-3H3,(H,29,33);1H3,(H,2,3,4)/t22-;/m1./s1

InChI key

DUGMCDWNXXFHDE-VZYDHVRKSA-N

Biochem/physiol Actions

MK-677 is a potent, non-peptide ghrelin receptor agonist.
It is also known as ibutamoren mesylate. MK-677 is a mimic of growth hormone (GH)-releasing peptide and is orally active.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library