Skip to Content
Merck
All Photos(1)

Key Documents

SML0947

Sigma-Aldrich

YN1

≥98% (HPLC)

Synonym(s):

7,8-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[o]1c2c([c](c(c1)c3ccc(cc3)O)=O)ccc(c2O)O

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-15-10(13(11)18)5-6-12(17)14(15)19/h1-7,16-17,19H

InChI key

BMZFZTMWBCFKSS-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

YN1 is a potent inhibitor of PFKFB3 (6-Phosphfructo-2-kinase/fructose-2,6-bisphosphatase: IC50 = 670 nM). PFKFB3 is over expressed in many cancers, and catalyzes the production of fructose-2,6-bisphosphate, which can potentiate PFK1 activity, enhancing glycolysis in tumor cells. YN1 inhibits glycolysis and proliferation in HeLa cells.
YN1 is a potent inhibitor of PFKFB3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Jun Tang et al.
Journal of inorganic biochemistry, 105(12), 1623-1629 (2011-11-11)
A new series of complexes of a ligand 4', 7, 8-trihydroxy-isoflavone with transition metal (zinc, copper, manganese, nickel, cobalt) and selenium have been synthesized and characterized with the aid of elemental analysis, IR, electron ionization mass spectrum (EI-MS) and (1)H
S Funayama et al.
The Journal of antibiotics, 42(9), 1350-1355 (1989-09-01)
Structures of three antioxidant isoflavonoids isolated from the cultured broth of Streptomyces sp. OH-1049 were shown to be 4',7,8-trihydroxyisoflavone (1), 3',4',7-trihydroxyisoflavone (2) and 8-chloro-3',4',5,7-tetrahydroxyisoflavone (3), respectively. Among them, 3 is a novel isoflavonoid possessing a chlorine atom in the molecule.
K Komiyama et al.
The Journal of antibiotics, 42(9), 1344-1349 (1989-09-01)
Three antioxidant isoflavonoids characterized as 4',7,8-trihydroxyisoflavone (1), 3',4',7-trihydroxyisoflavone (2) and 8-chloro-3',4',5,7-tetrahydroxyisoflavone (3) were isolated from the cultured broth of Streptomyces sp. OH-1049. Among them, 3 is a novel isoflavonoid possessing a chlorine atom in the molecule. In in vitro studies
Fabiola Diniz et al.
Nature communications, 14(1), 7733-7733 (2023-11-26)
Nephron endowment at birth impacts long-term renal and cardiovascular health, and it is contingent on the nephron progenitor cell (NPC) pool. Glycolysis modulation is essential for determining NPC fate, but the underlying mechanism is unclear. Combining RNA sequencing and quantitative

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service