Skip to Content
Merck
All Photos(1)

Key Documents

SML0942

Sigma-Aldrich

GSK2193874

≥98% (HPLC)

Synonym(s):

3-([1,4′-Bipiperidin]-1′-ylmethyl)-7-bromo-N-(1-phenylcyclopropyl)-2-[3-(trifluoromethyl)phenyl]-4-quinolinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H38BrF3N4O
CAS Number:
Molecular Weight:
691.62
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL, clear

storage temp.

room temp

SMILES string

FC(F)(F)c1cc(ccc1)c2nc3c(c(c2CN6CCC(CC6)N7CCCCC7)C(=O)NC5(CC5)c4ccccc4)ccc(c3)Br

InChI

1S/C37H38BrF3N4O/c38-28-12-13-30-32(23-28)42-34(25-8-7-11-27(22-25)37(39,40)41)31(24-44-20-14-29(15-21-44)45-18-5-2-6-19-45)33(30)35(46)43-36(16-17-36)26-9-3-1-4-10-26/h1,3-4,7-13,22-23,29H,2,5-6,14-21,24H2,(H,43,46)

InChI key

UIVOZBSCHXCGPS-UHFFFAOYSA-N

Application

GSK2193874 has been used as a transient receptor potential vanilloid 4 (TRPV4) antagonist:
  • to study its effects on GSK101-induced colon contractions in mice
  • to study its effects on murine compact bone-derived osteoblasts(CB-OB)
  • to study its effects on lung injury post-lipopolysaccharide (LPS) in mice

Biochem/physiol Actions

GSK2193874 is a very potent, specific antagonist of TRPV4 ion channels (IC50 = 50 nM). Lung edema caused by high pulmonary venous pressure (PVP) is driven by TRPV4 activity. GSK2193874 blocks TRPV4-mediated calcium influx in cells expressing native and recombinant TRPV4, and inhibits vascular permeability and lung edema in isolated rodent and canine lungs subjected to high PVP. The compound also resolves pulmonary edema in murine myocardial infarction model.
GSK2193874 is a very potent, specific antagonist of TRPV4.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheikh Rayees et al.
Cell reports, 27(3), 793-805 (2019-04-18)
Alveolar macrophages (AMs), upon sensing pathogens, trigger host defense by activating toll-like receptor 4 (TLR4), but the counterbalancing mechanisms that deactivate AM inflammatory signaling and prevent lethal edema, the hallmark of acute lung injury (ALI), remain unknown. Here, we demonstrate
Jialie Luo et al.
Immunity, 49(1), 107-119 (2018-07-01)
Intestinal macrophages are critical for gastrointestinal (GI) homeostasis, but our understanding of their role in regulating intestinal motility is incomplete. Here, we report that CX3C chemokine receptor 1-expressing muscularis macrophages (MMs) were required to maintain normal GI motility. MMs expressed the
Tomasz Janczi et al.
Cells, 10(10) (2021-10-24)
Mechanotransduction is elicited in cells upon the perception of physical forces transmitted via the extracellular matrix in their surroundings and results in signaling events that impact cellular functions. This physiological process is a prerequisite for maintaining the integrity of diarthrodial
Elena Cambria et al.
Cells, 9(7) (2020-07-28)
Mechanical loading and inflammation interact to cause degenerative disc disease and low back pain (LBP). However, the underlying mechanosensing and mechanotransductive pathways are poorly understood. This results in untargeted pharmacological treatments that do not take the mechanical aspect of LBP
Nicholas Mikolajewicz et al.
eLife, 7 (2018-10-17)
Bone cells sense and actively adapt to physical perturbations to prevent critical damage. ATP release is among the earliest cellular responses to mechanical stimulation. Mechanical stimulation of a single murine osteoblast led to the release of 70 ± 24 amole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service