Skip to Content
Merck

Skip To

SML0906

HPOB

≥98% (HPLC)

Synonym(s):

4-[(Hydroxyamino)carbonyl]-N-(2-hydroxyethyl)-N-phenyl-benzeneacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H18N2O4
CAS Number:
Molecular Weight:
314.34
UNSPSC Code:
12352202
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

N(O)C(=O)c1ccc(cc1)CC(=O)N(CCO)c2ccccc2

InChI

1S/C17H18N2O4/c20-11-10-19(15-4-2-1-3-5-15)16(21)12-13-6-8-14(9-7-13)17(22)18-23/h1-9,20,23H,10-12H2,(H,18,22)

InChI key

RFAZNTABYJYOAR-UHFFFAOYSA-N

Biochem/physiol Actions

HPOB is a very potent, selective inhibitor of HDAC6.
HPOB is a very potent, selective inhibitor of HDAC6. HPOB inhibits proliferation of normal and transformed cell lines but does not induce cell death. The compound HPOB potentiates the effects of DNA-damaging cytotoxic anti-cancer drugs.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters