Skip to Content

SML0882

Aurora-A Inhibitor I

≥98% (HPLC)

Synonym(s):

TC‐S7010, N-(2-Chlorophenyl)-4-(2-(4-(2-(4-ethylpiperazin-1-yl)-2-oxoethyl)phenylamino)-5-fluoropyrimidin-4-ylamino)benzamide, N-(2-Chlorophenyl)-4-[[2-[[4-[2-(4-ethyl-1-piperazinyl)-2-oxoethyl]phenyl]amino]-5-fluoro-4-pyrimidinyl]amino]-benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H31ClFN7O2
CAS Number:
Molecular Weight:
588.07
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(nc(nc1)Nc4ccc(cc4)CC(=O)N5CCN(CC5)CC)Nc2ccc(cc2)C(=O)Nc3c(cccc3)Cl

InChI

1S/C31H31ClFN7O2/c1-2-39-15-17-40(18-16-39)28(41)19-21-7-11-24(12-8-21)36-31-34-20-26(33)29(38-31)35-23-13-9-22(10-14-23)30(42)37-27-6-4-3-5-25(27)32/h3-14,20H,2,15-19H2,1H3,(H,37,42)(H2,34,35,36,38)

InChI key

AKSIZPIFQAYJGF-UHFFFAOYSA-N

Application

Aurora-A Inhibitor I has been used to study its effect on cell viability of human megakaryoblastic cell line (SET-2) and human erythroleukemia cell line (HEL) by methylthiazoletetrazolium (MTT) assay.

Biochem/physiol Actions

Aurora-A Inhibitor I is a potent inhibitor of Aurora kinase A with an IC50 of 3.4 nM and 1000-fold selectivity for Aurora A over Aurora B (better than VX-680) and even higher for CDKs. Antiproliferative IC50 values were in the vicinity of 1 μM against the human colorectal cancer cell line HCT116.
Aurora-A Inhibitor I is a potent inhibitor of Aurora kinase A.
Aurora-A Inhibitor I, also known as TC‐S7010, stimulates G2/M cell cycle arrest and apoptosis in ARID1A −/− cells.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library