Skip to Content
Merck
All Photos(1)

Key Documents

SML0881

Sigma-Aldrich

Rivastigmine tartrate

≥98% (HPLC)

Synonym(s):

ENA-713, Ethylmethyl-carbamic acid 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, N-Ethyl-N-methyl-carbamic acid 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester tartrate, S-Rivastigmine tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O2 · C4H6O6
CAS Number:
Molecular Weight:
400.42
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

N([C@@H](C)c1cc(ccc1)OC(=O)N(CC)C)(C)C.O[C@@H]([C@H](O)C(=O)O)C(=O)O

InChI

1S/C14H22N2O2.C4H6O6/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4;5-1(3(7)8)2(6)4(9)10/h7-11H,6H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t11-;1-,2-/m00/s1

InChI key

GWHQHAUAXRMMOT-RWALOXMOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rivastigmine tartrate is a cholinesterase inhibitor. It is also called as exelon.

Application

Rivastigmine tartrate has been used in cholinesterase inhibition assay.

Biochem/physiol Actions

Rivastigmine is an orally available, brain penetrant, reversible cholinesterase inhibitor that enhances cognitive function in patients with Alzheimer′s and Parkinson′s diseases. Rivastigmine inhibits both butyrylcholinesterase and acetylcholinesterase.
Rivastigmine is an orally available, brain penetrant, reversible cholinesterase inhibitor.
Rivastigmine tartrate is used to treat Alzheimer′s disease. It helps to improve thinking ability. It helps to increase cholinergic function by inhibiting acetylcholinesterase.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Synthesis and Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 4-Disubstituted quinazolines as amyloid-β aggregation inhibitors with dual cholinesterase inhibition and antioxidant properties: Development and structure-activity relationship (SAR) studies.
Mohamed T and Rao PPN
European Journal of Medicinal Chemistry, 126, 823-843 (2017)
Desk Reference of Clinical Pharmacology, Second Edition, 824-824 (2007)
The Complete Pill Guide, 1151-1151 (2003)
Jacqueline Birks et al.
The Cochrane database of systematic reviews, 5(5), CD004744-CD004744 (2013-06-04)
Vascular dementia represents the second most common type of dementia after Alzheimer's disease. In older patients, in particular, the combination of vascular dementia and Alzheimer's disease is common, and is referred to as mixed dementia. The classification of vascular dementia
Kevin R Peters
Journal of the American Geriatrics Society, 61(7), 1170-1174 (2013-05-29)
To highlight the utility of using an effect size analysis to communicate the effectiveness of treatment interventions. Secondary analysis. Previously published systematic review on cholinesterase inhibitors (ChEIs) in Alzheimer's disease. Individuals with mild to moderate Alzheimer's disease. Six-month randomized controlled

Articles

Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources.

Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources.

Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources.

Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service