Skip to Content
Merck

SML0832

TLR4-IN-C34

≥98% (HPLC), toll-like receptor 4TLR4 inhibitor, powder

Synonym(s):

2-Acetamidopyranoside, C34, Isopropyl 2-(acetylamino)-2-deoxy α-D-glucopyranoside 3,4,6-triacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H27NO9
CAS Number:
Molecular Weight:
389.40
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

TLR4-IN-C34, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

N([C@H]1[C@H](O[C@@H]([C@H]([C@@H]1OC(=O)C)OC(=O)C)COC(=O)C)OC(C)C)C(=O)C

InChI

1S/C17H27NO9/c1-8(2)24-17-14(18-9(3)19)16(26-12(6)22)15(25-11(5)21)13(27-17)7-23-10(4)20/h8,13-17H,7H2,1-6H3,(H,18,19)/t13-,14-,15-,16-,17+/m1/s1

InChI key

KMIQMFHPUJUDMC-HHARLNAUSA-N

General description

TLR4-IN-C34 is an inhibitor of toll-like receptor 4 (TLR4) in vitro and in vivo.

Application

TLR4-IN-C34 has been used as an inhibitor in the HFD-feeding mice to verify the aforementioned in vitro pathway (stearic acid/LDH-a/cytokines) in vivo.

Biochem/physiol Actions

TLR4-IN-C34 is a potent and specific inhibitor of toll-like receptor 4 (TLR4), which inhibits TLR4 signaling in vivo and in vitro. TLR4-IN-C34, also known as C34, is an aminomonosaccharide that inhibits TLR4 signaling by docking with the hydrophobic pocket of the TLR4 co-receptor, myeloid differentiation protein-2 (MD-2). C34 has been shown to diminish the severity of experimental necrotizing enterocolitis (NEC).
TLR4-IN-C34 is a potent and specific inhibitor of toll-like receptor 4 (TLR4).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library