Skip to Content
Merck
All Photos(1)

Key Documents

SML0768

Sigma-Aldrich

GSK2801

≥98% (HPLC)

Synonym(s):

1-(7-(2-Methylsulfonyl-phenyl)-4-propoxy-1-aza-bicyclo[4.3.0]nona-2,4,6,8-tetraen-9-yl)-ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO4S
CAS Number:
Molecular Weight:
371.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

, white to pink to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCOC1=CC2=C(C3=CC=CC=C3S(=O)(C)=O)C=C(C(C)=O)N2C=C1

InChI

1S/C20H21NO4S/c1-4-11-25-15-9-10-21-18(14(2)22)13-17(19(21)12-15)16-7-5-6-8-20(16)26(3,23)24/h5-10,12-13H,4,11H2,1-3H3

InChI key

KHWCPNJRJCNVRI-UHFFFAOYSA-N

Biochem/physiol Actions

GSK2801 is a potent inhibitor of the BAZ2 family of bromodomain containing proteins. BAZ2A is an essential component of the nucleolar remodeling complex (NoRC), which mediates recruitment of histone modifying enzymes and DNA methylase involved in the silencing of ribosomal RNA transcription. BAZ2B is believed to be involved in regulating nucleosome mobilization along linear DNA. For full characterization details, please visit the GSK2801 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

GSK2801 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

GSK2801 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the GSK2801 probe summary on the Chemical Probes Portal website.
Light sensitive

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natali Abeywickrama-Samarakoon et al.
Nature communications, 11(1), 419-419 (2020-01-23)
Hepatitis Delta virus (HDV) is a satellite of Hepatitis B virus with a single-stranded circular RNA genome. HDV RNA genome synthesis is carried out in infected cells by cellular RNA polymerases with the assistance of the small hepatitis delta antigen
Nathaniel W Mabe et al.
Cell reports, 33(5), 108341-108341 (2020-11-05)
Dysregulated gene expression is a common feature of cancer and may underlie some aspects of tumor progression, including tumor relapse. Here, we show that recurrent mammary tumors exhibit global changes in gene expression and histone modifications and acquire dependence on

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service