Skip to Content
Merck
All Photos(1)

Key Documents

SML0748

Sigma-Aldrich

Carbetocin acetate

≥95% (HPLC)

Synonym(s):

N-(4-Mercapto-1-oxobutyl)-O-methyl-L-tyrosyl-L-isoleucyl-L-glutaminyl-L-asparaginyl-L-cysteinyl-L-prolyl-L-leucyl-glycinamidecyclic (1-5)-thioether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H69N11O12S · xC2H4O2
CAS Number:
Molecular Weight:
988.16 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

storage temp.

−20°C

SMILES string

S1C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CCC1)Cc3ccc(cc3)OC)[C@H](CC)C)CCC(=O)N)CC(=O)N)C(=O)N2[C@@H](CCC2)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N

InChI

1S/C45H69N11O12S/c1-6-25(4)38-44(66)51-28(15-16-34(46)57)40(62)52-31(21-35(47)58)41(63)54-32(23-69-18-8-10-37(60)50-30(42(64)55-38)20-26-11-13-27(68-5)14-12-26)45(67)56-17-7-9-33(56)43(65)53-29(19-24(2)3)39(61)49-22-36(48)59/h11-14,24-25,28-33,38H,6-10,15-23H2,1-5H3,(H2,46,57)(H2,47,58)(H2,48,59)(H,49,61)(H,50,60)(H,51,66)(H,52,62)(H,53,65)(H,54,63)(H,55,64)/t25-,28-,29-,30-,31-,32-,33-,38-/m0/s1

InChI key

NSTRIRCPWQHTIA-DTRKZRJBSA-N

Biochem/physiol Actions

Carbetocin is a potent agonist of the oxytocin receptor, with improved in vivo stability over oxytocin.
Carbetocin is a potent agonist of the oxytocin receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas von Erlach et al.
Nature biomedical engineering, 4(5), 544-559 (2020-04-29)
Monolayers of cancer-derived cell lines are widely used in the modelling of the gastrointestinal (GI) absorption of drugs and in oral drug development. However, they do not generally predict drug absorption in vivo. Here, we report a robotically handled system
Barbara Chruścicka et al.
ACS chemical neuroscience, 10(7), 3225-3240 (2019-05-01)
The oxytocin receptor (OTR) and the 5-hydroxytryptamine 2A receptor (5-HTR2A) are expressed in similar brain regions modulating central pathways critical for social and cognition-related behaviors. Signaling crosstalk between their endogenous ligands, oxytocin (OT) and serotonin (5-hydroxytryptamine, 5-HT), highlights the complex
Shauna E Wallace Fitzsimons et al.
Neuropharmacology, 152, 90-101 (2018-12-26)
Oxytocin mediates its behavioural effects via the centrally expressed oxytocin receptor (OTR). Oxytocin signalling has been implicated in multiple disorders involving centrally regulated pathways, including obesity, autism, schizophrenia and depression. The OTR has been described to have a complex downstream
A secondary analysis of a randomized placebo-controlled trial comparing the analgesic effects of oxytocin with carbetocin: postcesarean delivery morphine equivalents.
Ewa Gawecka et al.
Anesthesia and analgesia, 119(4), 1004-1004 (2014-09-19)

Articles

Sigma-Aldrich offers many products related to vasopressin and oxytocin receptors for your research needs.

Sigma-Aldrich offers many products related to vasopressin and oxytocin receptors for your research needs.

Sigma-Aldrich offers many products related to vasopressin and oxytocin receptors for your research needs.

Sigma-Aldrich offers many products related to vasopressin and oxytocin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service