Skip to Content
Merck
All Photos(1)

Key Documents

SML0709

Sigma-Aldrich

GSK-J1

≥98% (HPLC)

Synonym(s):

3-((6-(4,5-Dihydro-1H-benzo[d]azepin-3(2H)-yl)-2-(pyridin-2-yl)pyrimidin-4-yl)amino)propanoic acid; N-[2-(2-pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-Alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N5O2
CAS Number:
Molecular Weight:
389.45
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(CCNC1=CC(N2CCC(C=CC=C3)=C3CC2)=NC(C4=CC=CC=N4)=N1)=O

InChI

1S/C22H23N5O2/c28-21(29)8-12-24-19-15-20(26-22(25-19)18-7-3-4-11-23-18)27-13-9-16-5-1-2-6-17(16)10-14-27/h1-7,11,15H,8-10,12-14H2,(H,28,29)(H,24,25,26)

InChI key

AVZCPICCWKMZDT-UHFFFAOYSA-N

Application

GSK-J1 has been used in formaldehyde dehydrogenase (FDH)-coupled demethylase assay.

Biochem/physiol Actions

GSK-J1 is a potent selective jumonji H3K27 demethylase inhibitor. Jumonji C domain-containing histone demethylases (JHDMs) are Fe(II) and α-ketoglutarate dependent enzymes that oxygenate methylated histone lysine residues and thereby cause their demethylation. GSK-J1 is selective for the KDM6 subfamily members JMJD3 and UTX with an IC50 of 60 nM in a JMJD3 assay, and is inactive against other demethylases of the JMJ family and over 100 tested kinases and histone deacetylases. For full characterization details, please visit the GSK-J1 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
GSK-J1 is also termed as 3-((6-(4,5-Dihydro-1H-benzo[d]azepin-3(2H)-yl)-2-(pyridin-2-yl)pyrimidin-4-yl)amino)propanoate. It may disturb the differentiation of specific neuronal subtypes in growing rat retina.

Features and Benefits

GSK-J1 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small Molecule GSK-J1 Affects Differentiation of Specific Neuronal Subtypes in Developing Rat Retina.
Raeisossadati R, et al.
Molecular Neurobiology, 1-12 (2018)
The Inhibitor Index: A Desk Reference on Enzyme Inhibitors, Receptor Antagonists, Drugs, Toxins, Poisons, Biologics, and Therapeutic Leads, 1362-1362 (2017)
Christopher E Gibson et al.
Hormone research in paediatrics, 89(6), 413-422 (2018-06-15)
Previous case reports have suggested a possible association of congenital hyperinsulinism with Turner syndrome. We examined the clinical and molecular features in girls with both congenital hyperinsulinism and Turner syndrome seen at The Children's Hospital of Philadelphia (CHOP) between 1974
Characterization of a linked Jumonji domain of the KDM5/JARID1 family of histone H3 lysine 4 demethylases.
Horton JR, et al.
The Journal of Biological Chemistry, 291(6), 2631-2646 (2016)
Reza Raeisossadati et al.
Molecular neurobiology, 56(3), 1972-1983 (2018-07-08)
Histone post-translational modification has been shown to play a pivotal role in regulating gene expression and fate determination during the development of the central nervous system. Application of pharmacological blockers that control histone methylation status has been considered a promising

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service