Skip to Content
Merck
All Photos(1)

Key Documents

SML0694

Sigma-Aldrich

BX-795 hydrochloride

≥98% (HPLC)

Synonym(s):

N-[3-[[5-iodo-4-[[3-[(2-thienylcarbonyl)amino]propyl]amino]-2-pyrimidinyl]amino]phenyl]-1-Pyrrolidinecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26IN7O2S · xHCl
CAS Number:
Molecular Weight:
591.47 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.Ic1cnc(Nc2cccc(NC(=O)N3CCCC3)c2)nc1NCCCNC(=O)c4cccs4

InChI

1S/C23H26IN7O2S/c24-18-15-27-22(30-20(18)25-9-5-10-26-21(32)19-8-4-13-34-19)28-16-6-3-7-17(14-16)29-23(33)31-11-1-2-12-31/h3-4,6-8,13-15H,1-2,5,9-12H2,(H,26,32)(H,29,33)(H2,25,27,28,30)

InChI key

VAVXGGRQQJZYBL-UHFFFAOYSA-N

Application

BX-795 hydrochloride has been used to study the effect of kinase inhibition on human endogenous retroviruses (HERVs) transcription activation.

Biochem/physiol Actions

BX-795 competes for the ATP (adenosine triphosphate) binding pocket of 3-phosphoinositide-dependent kinase-1 (PDK1) with its substrate ATP. In vitro assays reveal that BX-795 might inhibit Unc-51 (serine/threonine-protein kinase)-like autophagy activating kinase (ULK1).
BX-795 was initially characterized as a potent inhbitor of 3-phosphoinositide-dependent kinase-1. Additional studies with BX-795 have shown that the molecule is also a potent dual inhibitor of IKKe and TBK1 which are known to regulate the expression of interferons via the phosphorylation of interferon regulatory factor 3 (IRF3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compound signaling activates endogenous retroviruses by inducing enhancer and gene-neighborhood transcription.
Azebi S, et al.
bioRxiv, 284695-284695 (2018)
Discovery and structure of a new inhibitor scaffold of the autophagy initiating kinase ULK1.
Lazarus M B and Kevan M S
Bioorganic & Medicinal Chemistry, 23(17), 5483-5488 (2015)
Nagarjuna R Cheemarla et al.
medRxiv : the preprint server for health sciences (2021-02-04)
The interferon response is a potent antiviral defense mechanism, but its effectiveness depends on its timing relative to viral replication. Here, we report viral replication and host response kinetics in patients at the start of SARS-CoV-2 infection and explore the
Weijian Ye et al.
PLoS pathogens, 14(10), e1007298-e1007298 (2018-10-05)
Natural killer (NK) cells provide the first line of defense against malaria parasite infection. However, the molecular mechanisms through which NK cells are activated by parasites are largely unknown, so is the molecular basis underlying the variation in NK cell
Richard I Feldman et al.
The Journal of biological chemistry, 280(20), 19867-19874 (2005-03-18)
The phosphoinositide 3-kinase/3-phosphoinositide-dependent kinase 1 (PDK1)/Akt signaling pathway plays a key role in cancer cell growth, survival, and tumor angiogenesis and represents a promising target for anticancer drugs. Here, we describe three potent PDK1 inhibitors, BX-795, BX-912, and BX-320 (IC(50)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service