Skip to Content
Merck
All Photos(1)

Key Documents

SML0649

Sigma-Aldrich

Deshydroxy LY-411575

≥98% (HPLC)

Synonym(s):

γ-Secretase inhibitor XX, (S,S)-2-[2-(3,5-Difluorophenyl)acetylamino]-N-(5-methyl-6-oxo-6,7-dihydro-5H-dibenzo[b,d]azepin-7-yl)propionamide, DBZ, Dibenzazepine, GSI-XX, N-[(1S)-2-[[(7S)-6,7-Dihydro-5-methyl-6-oxo-5H-dibenz[b,d]azepin-7-yl]amino]-1-methyl-2-oxoethyl]-3,5-difluoro-benzeneacetamide, YO-01027

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H23F2N3O3
CAS Number:
Molecular Weight:
463.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -128.0 to -154.0°-0.25 in methanol

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

Fc1cc(cc(c1)CC(=O)N[C@@H](C)C(=O)N[C@H]2c3c(cccc3)c4c(cccc4)N(C2=O)C)F

InChI

1S/C26H23F2N3O3/c1-15(29-23(32)13-16-11-17(27)14-18(28)12-16)25(33)30-24-21-9-4-3-7-19(21)20-8-5-6-10-22(20)31(2)26(24)34/h3-12,14-15,24H,13H2,1-2H3,(H,29,32)(H,30,33)/t15-,24-/m0/s1

InChI key

QSHGISMANBKLQL-OWJWWREXSA-N

Application

Deshydroxy LY-411575 has been used in dibenzazepine (DBZ) treatment and chemical treatment.

Biochem/physiol Actions

Deshydroxy LY-411575 is a cell-permeable γ-secretase inhibitor and inhibtor of Notch processing.
Deshydroxy LY-411575 is a cell-permeable γ-secretase inhibitor and inhibtor of Notch processing. Deshydroxy LY-411575 significantly lowers both brain and plasma Aβ40 levels by ~72% in Tg2576 mutant APP transgenic mouse model and potently blocks Notch processing, with an IC50 of 1.7 nM in SupT1 cells.
LY411575 prevents SPP (signal peptide peptidase) dependent cleavage of HCV (hepatitis C virus) core protein.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pooja Hor et al.
iScience, 23(5), 101083-101083 (2020-05-08)
Expansion of pulmonary neuroendocrine cells (PNECs) is a pathological feature of many human lung diseases. Human PNECs are inherently difficult to study due to their rarity (<1% of total lung cells) and a lack of established protocols for their isolation.
G protein-coupled receptor 183 facilitates endothelial-to-hematopoietic transition via Notch1 inhibition.
Zhang P, et al.
Cell Research, 25(10), 1093-1093 (2015)
Yan Li et al.
Development (Cambridge, England), 149(10) (2022-05-04)
The earliest hematopoietic stem and progenitor cells (HSPCs) are generated from the ventral wall of the dorsal aorta, through endothelial-to-hematopoietic transition during vertebrate embryogenesis. Notch signaling is crucial for HSPC generation across vertebrates; however, the precise control of Notch during
Qiuping He et al.
Experimental hematology, 51, 1-6 (2017-05-01)
During development, hematopoietic stem cells (HSCs) undergo a rapid expansion in the fetal liver (FL) after their emergence in the aorta-gonad-mesonephros (AGM) region. We recently reported that the endolysosomal trafficking factor BLOS2, encoded by the Bloc1s2 gene, regulates HSC/hematopoietic progenitor
Inhibitory effect of presenilin inhibitor LY411575 on maturation of hepatitis C virus core protein, production of the viral particle and expression of host proteins involved in pathogenicity.
Otoguro T, et al.
Microbiology and Immunology, 60(11), 740-753 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service