Skip to Content
Merck
All Photos(1)

Key Documents

SML0648

Sigma-Aldrich

CHR-6494 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

3-(1H-Indazol-5-yl)-N-propyl-imidazo[1,2-b]pyridazin-6-amine trifluoroacetate salt, 3-(1H-Indazol-5-yl)-N-propylimidazo[1,2-b]pyridazin-6-amine trifluoroacetate salt, CHR 6494 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N6 · CF3COOH
CAS Number:
Molecular Weight:
406.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.CCCNc1ccc2ncc(-c3ccc4[nH]ncc4c3)n2n1

InChI

1S/C16H16N6.C2HF3O2/c1-2-7-17-15-5-6-16-18-10-14(22(16)21-15)11-3-4-13-12(8-11)9-19-20-13;3-2(4,5)1(6)7/h3-6,8-10H,2,7H2,1H3,(H,17,21)(H,19,20);(H,6,7)

InChI key

ILWYDZNXJQESDI-UHFFFAOYSA-N

Biochem/physiol Actions

CHR-6494 is a potent and selective Haspin (Haploid Germ Cell-Specific Nuclear Protein Kinase) inhibitor that blocks H3T3ph phosphorylation. CHR-6494 causes cell cycle arrest in G2/M and subsequent apoptosis. CHR-6494 causes an abnormal duplication of centrosomes and mitotic catastrophe in cancer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Quadri et al.
iScience, 26(10), 108011-108011 (2023-10-16)
Throughout mitosis, a plethora of processes must be efficiently concerted to ensure cell proliferation and tissue functionality. The mitotic spindle does not only mediate chromosome segregation, but also defines the axis of cellular division, thus determining tissue morphology. Functional spindle
Structure Activity Relationship Studies around DB18, a Potent and Selective Inhibitor of CLK Kinases.
Brahmaiah, et al.
Molecules (Basel), 27 (2022)
Bruno Oyallon et al.
European journal of medicinal chemistry, 154, 101-109 (2018-05-21)
We identified a new series of quinoxaline-2-carboxylic acid derivatives, targeting the human proviral integration site for Moloney murine leukemia virus-1 (HsPim-1) kinase. Seventeen analogues were synthesized providing useful insight into structure-activity relationships studied. Docking studies realized in the ATP pocket
Marc-Antoine Bazin et al.
European journal of medicinal chemistry, 210, 112956-112956 (2020-11-06)
Leishmaniasis constitutes a severe public health problem, with an estimated prevalence of 12 million cases. This potentially fatal disease has a worldwide distribution and in 2012, the fatal Visceral Leishmaniasis (VL) was declared as new emerging disease in Europe, mainly
Roberto Quadri et al.
International journal of molecular sciences, 22(14) (2021-07-25)
Primary cilia are commonly found on most quiescent, terminally differentiated cells and play a major role in the regulation of the cell cycle, cell motility, sensing, and cell-cell communication. Alterations in ciliogenesis and cilia maintenance are causative of several human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service