Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C25H26N2O5
CAS Number:
Molecular Weight:
434.48
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 15 mg/mL, clear
storage temp.
2-8°C
SMILES string
N(CC4(CCOCC4)c3ccc(cc3)OC)C(=O)c1ccc(cc1)NC(=O)c2[o]ccc2
InChI
1S/C25H26N2O5/c1-30-21-10-6-19(7-11-21)25(12-15-31-16-13-25)17-26-23(28)18-4-8-20(9-5-18)27-24(29)22-3-2-14-32-22/h2-11,14H,12-13,15-17H2,1H3,(H,26,28)(H,27,29)
InChI key
ZJZWZIXSGNFWQQ-UHFFFAOYSA-N
Application
JW55 has been used as a tankyrase-specific inhibitor to study its effects on the transcriptional activity of SRY-related HMG-box gene 9 (Sox9) in chondrocytes.
Biochem/physiol Actions
JW55 can decrease the formation of mouse colon adenoma in vivo. It possesses a methoxyphenyl moiety spreading towards the G-loop.
JW55 is a potent and selective inhibitor of Wnt/β-catenin signaling, that functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2
JW55 is a potent and selective inhibitor of Wnt/β-catenin signaling, that functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2) leading to stabilization of AXIN2 followed by increased degradation of β-catenin. JW55 specifically inhibits canonical Wnt signaling that results in reduced cell-cycle progression, proliferation, and colony formation in colon carcinoma cell lines.
Other Notes
JW55 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the JW55 probe summary on the Chemical Probes Portal website.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.