Skip to Content
Merck
All Photos(1)

Key Documents

SML0613

Sigma-Aldrich

Rofecoxib

≥98% (HPLC)

Synonym(s):

4-[4-(Methylsulfonyl)phenyl]-3-phenyl-2(5H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O4S
CAS Number:
Molecular Weight:
314.36
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(C)c1ccc(cc1)C2=C(C(=O)OC2)c3ccccc3

InChI

1S/C17H14O4S/c1-22(19,20)14-9-7-12(8-10-14)15-11-21-17(18)16(15)13-5-3-2-4-6-13/h2-10H,11H2,1H3

InChI key

RZJQGNCSTQAWON-UHFFFAOYSA-N

Related Categories

Application

Rofecoxib has been used in high performance bioaffinity chromatography.

Biochem/physiol Actions

Rofecoxib is a selective cyclooxygenase-2 (COX-2) inhibitor with >800-fold COX-2 selectivity in CHO cells expressing human COX-1 and COX-2.
Rofecoxib is a selective cyclooxygenase-2 (COX-2) inhibitor.
Rofecoxib is derived from furanone and has the ability to cross human placenta. Along with anti-inflammatory action, it possesses analgesic and antipyretic properties. Cytosolic hepatic enzymes are responsible for the metabolism of rofecoxib. It is known to cause oligohydramnios and ductus arteriosus constrictions. Rofecoxib inhibits the action of CYP1A2 (cytochrome P450 family 1 subfamily A member 2). It might be associated with aseptic meningitis. Rofecoxib is known to ameliorate the risk of colorectal adenoma, but might contribute to toxicity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Kerstin Rabausch et al.
Circulation research, 96(1), e1-e6 (2004-12-14)
There is concern that cyclooxygenase (COX)-2 inhibitors may promote atherothrombosis by inhibiting vascular formation of prostacyclin (PGI2) and an increased thrombotic risk of COX-2 inhibitors has been reported. It is widely accepted that the prothrombotic effects of COX-2 inhibitors can
Peter Pyrko et al.
Molecular cancer, 5, 19-19 (2006-05-19)
2,5-Dimethyl-celecoxib (DMC) is a close structural analog of the selective cyclooxygenase-2 (COX-2) inhibitor celecoxib (Celebrex) that lacks COX-2-inhibitory function. However, despite its inability to block COX-2 activity, DMC is able to potently mimic the anti-tumor effects of celecoxib in vitro
J Patrick O'Connor et al.
Acta orthopaedica, 80(5), 597-605 (2009-11-18)
Non-steroidal anti-inflammatory drugs (NSAIDs) inhibit cyclooxygenase (COX) activity, which is the rate-limiting enzyme in the synthesis of prostaglandins. Previous studies have indicated that NSAID therapy, and in particular NSAIDs that specifically target the inflammatory cyclooxygenase (COX-2), impair bone healing. We
Rosivaldo S Borges et al.
Molecules (Basel, Switzerland), 24(1) (2019-01-06)
A drug design for safer phenylbutazone was been explored by reactivity and docking studies involving single electron transfer mechanism, as well as toxicological predictions. Several approaches about its structural properties were performed through quantum chemistry calculations at the B3LYP level
Krista F Huybrechts et al.
Medical care, 55(6), 545-551 (2017-05-16)
Many countries lack fully functional pharmacovigilance programs, and public budgets allocated to pharmacovigilance in industrialized countries remain low due to resource constraints and competing priorities. Using 3 case examples, we sought to estimate the public health and economic benefits resulting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service