Skip to Content
Merck
All Photos(1)

Key Documents

SML0564

Sigma-Aldrich

PD153035 hydrochloride

≥98% (HPLC)

Synonym(s):

4-[(3-Bromophenyl)amino]-6,7-dimethoxyquinazoline hydrochloride, AG 1517, PD 153035, SU 5271, ZM 252868

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14BrN3O2 · HCl
CAS Number:
Molecular Weight:
396.67
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Cl.COc1cc2ncnc(Nc3cccc(Br)c3)c2cc1OC

InChI

1S/C16H14BrN3O2.ClH/c1-21-14-7-12-13(8-15(14)22-2)18-9-19-16(12)20-11-5-3-4-10(17)6-11;/h3-9H,1-2H3,(H,18,19,20);1H

InChI key

ZJOKWAWPAPMNIM-UHFFFAOYSA-N

Biochem/physiol Actions

PD153035 is apotent and selective ATP competitive inhibitor of the epidermal growth factor receptor tyrosine kinase EGFR.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the EGFR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rainelli B Koumangoye et al.
Molecular cancer, 12(1), 167-167 (2013-12-21)
The expression of annexin A6 (AnxA6) in AnxA6-deficient non-invasive tumor cells has been shown to terminate epidermal growth factor receptor (EGFR) activation and downstream signaling. However, as a scaffolding protein, AnxA6 may stabilize activated cell-surface receptors to promote cellular processes
Amelia G Pérez et al.
Oncology reports, 44(4), 1649-1661 (2020-09-19)
Changes in protein levels in different components of the apical junctional complex occur in colorectal cancer (CRC). Claudin‑3 is one of the main constituents of tight junctions, and its overexpression can increase the paracellular flux of macromolecules, as well as
Naoya Hino et al.
Developmental cell, 53(6), 646-660 (2020-06-05)
During collective migration of epithelial cells, the migration direction is aligned over a tissue-scale expanse. Although the collective cell migration is known to be directed by mechanical forces transmitted via cell-cell junctions, it remains elusive how the intercellular force transmission
Shigeki Suzuki et al.
Molecular and cellular biochemistry, 476(4), 1673-1690 (2021-01-10)
Accumulating evidence suggests that specific non-coding RNAs exist in many types of malignant tissues, and are involved in cancer invasion and metastasis. However, little is known about the precise roles of non-coding RNAs in squamous cell carcinoma (SQCC) invasion and
P Dhawan et al.
Oncogene, 30(29), 3234-3247 (2011-03-09)
Claudin-2 is a unique member of the claudin family of transmembrane proteins, as its expression is restricted to the leaky epithelium in vivo and correlates with epithelial leakiness in vitro. However, recent evidence suggests potential functions of claudin-2 that are

Articles

Our company offers many products related to EGFR for your research needs.

Our company offers many products related to EGFR for your research needs.

Our company offers many products related to EGFR for your research needs.

Our company offers many products related to EGFR for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service