Skip to Content
Merck
All Photos(1)

Key Documents

SML0496

Sigma-Aldrich

CORM-3

Synonym(s):

Carbon monoxide releasing molecule 3, Tricarbonylchloro(glycinato)ruthenium (II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4ClNO5Ru
CAS Number:
Molecular Weight:
294.61
UNSPSC Code:
12352200
NACRES:
NA.77

form

powder

Quality Level

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Ru+2].[Cl-].NCC(=O)[O-].O=[C].O=[C].O=[C]

InChI

1S/C2H5NO2.3CO.ClH.Ru/c3-1-2(4)5;3*1-2;;/h1,3H2,(H,4,5);;;;1H;/q;;;;;+2/p-2

InChI key

BICHHHDSEZXKNN-UHFFFAOYSA-L

Application

CORM-3 (carbon monoxide (CO) releasing molecule-3) has been used to study its effect on NLRP3 (leucine-rich-repeat-containing receptor, pyrin-domain-containing 3) inflammasome activation via glycolysis in macrophages and also on hyperglycemia-induced IL-1β (interleukin-1 β) production. It has also been used to study its protective function against H2O2-induced apoptosis using primary rabbit lens epithelial cells.

Biochem/physiol Actions

CO possesses anti apoptotic function and offers protection against oxidative damage, promoting endothelial healing. CORM-3 is known to have therapeutic effects in transplantation, myocardial infarction and rheumatoid arthritis.
CORM-3 is a water-soluble carbon monoxide (CO) releasing molecule that can be used to study the effects of CO on cellular systems. Carbon monoxide (CO), produced during the degradation of heme by the enzyme heme oxygenase, has recently been found to be an important gaseous signaling mediator in mammalian cells CORM-3 has been shown to have anti-inflammatory and cardioprotective activity.
CORM-3 is a water-soluble carbon monoxide releasing molecule.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Carbon monoxide regulates glycolysis-dependent NLRP3 inflammasome activation in macrophages.
young S H, et al.
Biochemical and Biophysical Research Communications, 493(2), 957-963 (2017)
Carbon monoxide (CO) inhibits hydrogen peroxide (H2O2)?induced oxidative stress and the activation of NF-?B signaling in lens epithelial cells.
Huang Y, et al.
Experimental Eye Research, 166, 29-39 (2018)
CORM-3 Reactivity toward Proteins: The Crystal Structure of a Ru (II) Dicarbonyl? Lysozyme Complex.
Santos-Silva T, et al.
Journal of the American Chemical Society, 133(5), 1192-1195 (2011)
Ricardo Coletti et al.
Journal of neuroendocrinology, 31(2), e12686-e12686 (2019-01-12)
Nitric oxide (NO) negatively modulates the secretion of vasopressin (AVP), oxytocin (OT) and atrial natriuretic peptide (ANP) induced by the increase in extracellular osmolality, whereas carbon monoxide (CO) and hydrogen sulphide (H2 S) act to potentiate it; however, little information
Eric K Patterson et al.
Experimental biology and medicine (Maywood, N.J.), 246(21), 2338-2345 (2021-07-23)
In sepsis-induced inflammation, polymorphonuclear neutrophils (PMNs) contribute to vascular dysfunction. The serine proteases proteinase 3 (PR3) and human leukocyte elastase (HLE) are abundant in PMNs and are released upon degranulation. While HLE's role in inflammation-induced endothelial dysfunction is well studied

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service