Skip to Content
Merck
All Photos(1)

Documents

SML0376

Sigma-Aldrich

CGS 9895

≥98% (HPLC)

Synonym(s):

2,5-Dihydro-2-(4-methoxyphenyl)-3H-pyrazolo[4,3-c]quinolin-3-one, NSC 373970

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N3O2
CAS Number:
Molecular Weight:
291.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

light yellow to light brown

solubility

DMSO: >2 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)N2N=C3C(=CNc4ccccc34)C2=O

InChI

1S/C17H13N3O2/c1-22-12-8-6-11(7-9-12)20-17(21)14-10-18-15-5-3-2-4-13(15)16(14)19-20/h2-10,18H,1H3

InChI key

KSKRJZMRHSNRBX-UHFFFAOYSA-N

Biochem/physiol Actions

CGS 9895 is a GABA antagonist that acts via the benzodiazepine binding site of ag containing GABA receptors. CGS 9895 is the only known compound that can specifically enhance GABA-induced currents in ab subunit containing receptors, and acts at the extracellular a1+b3- subunit interface.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Cooper et al.
Physiology & behavior, 41(3), 247-255 (1987-01-01)
Several categories of compounds active at benzodiazepine receptors (BZR) in the brain have been distinguished: agonists, antagonists and the novel category of inverse agonist. In terms of their effects on ingestional responses (e.g., food, saline and water consumption), agonists increase
H E Shannon et al.
European journal of pharmacology, 92(1-2), 155-157 (1983-08-19)
The diazepam-like agonist and diazepam antagonist properties of the pyrazoloquinoline benzodiazepine receptor ligands CGS8216, CGS9895 and CGS9896 were evaluated in rats trained to discriminate between saline and 1.0 mg/kg of diazepam in a two-choice, discrete-trial procedure. None of the three
K Takada et al.
Life sciences, 44(4), 289-299 (1989-01-01)
Rats were trained to respond under 3-min fixed-interval schedules of food presentation, and effects of the benzodiazepine-receptor ligands, flumazenil, 2-(4-methoxy-phenyl)-pyrazolo[4,3-c]quinolin-3(5H)-one (CGS 9895), 3-carbo-t-butoxy-beta-carboline (beta-CCtB), and beta-carboline-3-carboxylic acid ethyl ester (beta-CCE) were assessed before and after the induction of tolerance to
H E Shannon et al.
Pharmacology, biochemistry, and behavior, 23(2), 317-323 (1985-08-01)
The effects of diazepam and the pyrazoloquinoline benzodiazepine receptor ligands CGS8216, CGS9896, and CGS9895 on schedule-controlled responding were studied in dogs. Responding was maintained under a multiple fixed-interval (FI) 5-min fixed-ratio (FR) 30 response schedule of food presentation. Diazepam (PO)
K Wegelius et al.
European journal of pharmacology, 263(1-2), 141-147 (1994-09-22)
The effects of benzodiazepine receptor ligands with different intrinsic activity profiles were studied on voluntary ethanol consumption in the selectively bred alcohol-preferring AA (Alko, Alcohol) rat line, and compared to those of an opiate antagonist, naloxone, and a serotonin uptake

Articles

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service