Skip to Content
Merck

SML0243

ML169

≥98% (HPLC)

Synonym(s):

2-[[1-[(5-Bromo-2-fluorophenyl)methyl]-1H-indol-3-yl]sulfonyl]-N-(5-methyl-3-isoxazolyl)-acetamide, VU0405652

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H17BrFN3O4S
CAS Number:
Molecular Weight:
506.34
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

Cc1cc(NC(=O)CS(=O)(=O)c2cn(Cc3cc(Br)ccc3F)c4ccccc24)no1

InChI

1S/C21H17BrFN3O4S/c1-13-8-20(25-30-13)24-21(27)12-31(28,29)19-11-26(18-5-3-2-4-16(18)19)10-14-9-15(22)6-7-17(14)23/h2-9,11H,10,12H2,1H3,(H,24,25,27)

InChI key

RUYRNYRGPSAXTK-UHFFFAOYSA-N

Biochem/physiol Actions

ML169 is a positive allosteric modulator (PAM) of the muscarinic acetylcholine receptor M1 (CHRM1). ML169 does not cross react with other muscarinic receptors and is able to penetrate the blood brain barrier.
ML169 is a positive allosteric modulator of muscarinic acetylcholine receptor M1.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library