Skip to Content
Merck
All Photos(1)

Documents

SML0235

Sigma-Aldrich

CDPPB

≥98% (HPLC)

Synonym(s):

3-Cyano-N-1,3-diphenyl-1H-pyrazol-5-yl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16N4O
CAS Number:
Molecular Weight:
364.40
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL at warmed to 60 °C

storage temp.

2-8°C

SMILES string

O.O=C(Nc1cc(nn1-c2ccccc2)-c3ccccc3)c4cccc(c4)C#N

InChI

1S/C23H16N4O.H2O/c24-16-17-8-7-11-19(14-17)23(28)25-22-15-21(18-9-3-1-4-10-18)26-27(22)20-12-5-2-6-13-20;/h1-15H,(H,25,28);1H2

InChI key

GZHZEGYXLNXEBV-UHFFFAOYSA-N

Application

CDPPB may be used in mGluR5-mediated cell signaling studies.

Biochem/physiol Actions

Activation of metabotropic glutamate 5 receptor by CDPPB enhances the function of NMDA receptor and markers of neuronal plasticity. This improves recognition memory and cognition deficits in schizophrenia.
CDPPB is a glutamate metabotropic mGluR5 positive allosteric modulator.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason M Uslaner et al.
Neuropharmacology, 57(5-6), 531-538 (2009-07-25)
In the search for strategies to treat schizophrenia, attention has focused on enhancing NMDA receptor function. In vitro experiments show that metabotropic glutamate 5 receptor (mGluR5) activation enhances NMDA receptor activity, and in vivo experiments indicate that mGluR5 positive allosteric
Hyejin Lee et al.
PLoS biology, 17(6), e2005326-e2005326 (2019-06-06)
Netrin-G ligand-3 (NGL-3) is a postsynaptic adhesion molecule known to directly interact with the excitatory postsynaptic scaffolding protein postsynaptic density-95 (PSD-95) and trans-synaptically with leukocyte common antigen-related (LAR) family receptor tyrosine phosphatases to regulate presynaptic differentiation. Although NGL-3 has been
Christina Gobin et al.
Psychopharmacology, 237(1), 115-125 (2019-08-26)
Cocaine use disorder (CUD) remains difficult to treat with no FDA-approved medications to reduce relapse. Antagonism of metabotropic glutamate receptor 5 (mGlu5) has been demonstrated to decrease cocaine-seeking but may also further compromise cognitive function in long-term cocaine users. Here we
Patrick K McCamphill et al.
Science translational medicine, 12(544) (2020-05-22)
Fragile X syndrome is caused by FMR1 gene silencing and loss of the encoded fragile X mental retardation protein (FMRP), which binds to mRNA and regulates translation. Studies in the Fmr1-/y mouse model of fragile X syndrome indicate that aberrant
J G Doria et al.
British journal of pharmacology, 169(4), 909-921 (2013-03-16)
Huntington's disease (HD) is an autosomal dominant neurodegenerative disorder caused by a polyglutamine expansion in the huntingtin protein. We have previously demonstrated that the cell signalling of the metabotropic glutamate receptor 5 (mGluR5) is altered in a mouse model of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service