Skip to Content
Merck
All Photos(1)

Key Documents

SML0203

Sigma-Aldrich

iCRT14

≥98% (HPLC)

Synonym(s):

5-[[2,5-Dimethyl-1-(3-pyridinyl)-1H-pyrrol-3-yl]methylene]-3-phenyl-2,4-thiazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17N3O2S
CAS Number:
Molecular Weight:
375.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Cc1cc(\C=C2/SC(=O)N(c3ccccc3)C2=O)c(C)n1-c4cccnc4

InChI

1S/C21H17N3O2S/c1-14-11-16(15(2)23(14)18-9-6-10-22-13-18)12-19-20(25)24(21(26)27-19)17-7-4-3-5-8-17/h3-13H,1-2H3/b19-12-

InChI key

NCSHZXNGQYSKLR-UNOMPAQXSA-N

Application

iCRT14 may be used in Wnt /β-catenin-mediated cell signaling studies.

Biochem/physiol Actions

Wnt / beta-catenin pathway inhibitor
iCRT14 belongs to the thiazolidinedione class of β-catenin-responsive transcription inhibitors. It decreases the levels of Dishevelled protein and modulates the binding of T-cell factor (TCF) to DNA. It results in consistent decrease in reduction of cell proliferation and tumor growth in colon cancer cells.
iCRT14 is a Wnt / β-catenin pathway inhibitor. It is a potent inhibitor of Catenin Responsive Transcription (CRT) reporter genes, as well as endogenous gene targets. iCRT14 also disrupts β-catenin-TCF4 interaction in a dose dependent manner, and causes G0/G1 arrest in colon tumor lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Wang et al.
Nature communications, 7, 10633-10633 (2016-02-05)
Cancer stem cells (CSCs) contribute to tumour heterogeneity, therapy resistance and metastasis. However, the regulatory mechanisms of cancer cell stemness remain elusive. Here we identify PCNA-associated factor (PAF) as a key molecule that controls cancer cell stemness. PAF is highly
The S-G2 phase enriched beta-catenin/TCF complex ensures cell survival and cell cycle progression
Ding Y, et al.
Journal of Cell Science, jcs-146977 (2014)
Daiana S Sánchez et al.
Biochemical and biophysical research communications, 512(2), 170-175 (2019-03-19)
This work was aimed to determine the effect of 17β-estradiol (17βE) on cell proliferation in human renal tubular epithelial cells (HRTEC) isolated from kidneys from pediatric subjects, as well as the role of estrogen receptors involved in the 17βE proliferative
Sohee Jun et al.
Nature communications, 7, 10994-10994 (2016-03-25)
Despite the implication of Wnt signalling in radioresistance, the underlying mechanisms are unknown. Here we find that high Wnt signalling is associated with radioresistance in colorectal cancer (CRC) cells and intestinal stem cells (ISCs). We find that LIG4, a DNA
Shiori Aono et al.
International journal of molecular sciences, 20(13) (2019-07-11)
Remarkable upregulation of the NRF2 (NFE2L2)-related transcription factor NRF3 (NFE2L3) in several cancer tissues and its correlation with poor prognosis strongly suggest the physiological function of NRF3 in tumors. Indeed, we had recently uncovered the function of NRF3, which promotes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service