Skip to Content
Merck
All Photos(1)

Key Documents

SML0083

Sigma-Aldrich

GSK 1059615 sodium salt hydrate

Synonym(s):

(5Z)-5-[[4-(4-pyridinyl)-6-quinolinyl]methylene]-2,4-thiazolidinedione sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H10N3O2S·Na · xH2O
CAS Number:
Molecular Weight:
355.35 (anhydrous basis)
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.44

form

powder

color

yellow

solubility

H2O: ≥8 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

O.[Na+].O=C1[N-]C(=O)C(\S1)=C\c2ccc3nccc(-c4ccncc4)c3c2

InChI

1S/C18H11N3O2S.Na.H2O/c22-17-16(24-18(23)21-17)10-11-1-2-15-14(9-11)13(5-8-20-15)12-3-6-19-7-4-12;;/h1-10H,(H,21,22,23);;1H2/q;+1;/p-1/b16-10-;;

InChI key

KKSRFFGUIONCPS-FLPKAINGSA-M

Biochem/physiol Actions

GSK 1059615 is a phosphatidylinositol-3-kinases (PI3K) inhibitor. GSK 1059615 inhibits proliferation of breast cancer BT474 cells via four different mechanisms that include retinoblastoma 1 (RB1)-mediated cell cycle arrest, elevated forkhead box protein O1 (FOXO) signaling, deduced MYC and transferrin receptor (TFRC) signaling and decreased cellular metabolism. Additionally, GSK 1059615 can suppress cell proliferation by decreasing mitogen-activated protein kinase (MAPK) signaling and imparts sensitivity to phosphoinositide 3-kinase (PI3K) inhibitor in cells resistant to the protein kinase B or AKT inhibitor. GSK 1059615 functions as a druggable target to reduce radiation-induced apoptosis in NCCIT cells. GSK 1059615 has entered clinical trial in patients with solid tumors or lymphoma and refractory malignancies.
GSK 1059615 is a potent inhibitor of PI3 Kinase. (IC50 = 2 nM)

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal D Zellefrow et al.
Radiation research, 178(3), 150-159 (2012-07-04)
Currently, there is a serious absence of pharmaceutically attractive small molecules that mitigate the lethal effects of an accidental or intentional public exposure to toxic doses of ionizing radiation. Moreover, cellular systems that emulate the radiobiologically relevant cell populations and
Pixu Liu et al.
Nature reviews. Drug discovery, 8(8), 627-644 (2009-08-01)
The phosphoinositide 3-kinase (PI3K) pathway is a key signal transduction system that links oncogenes and multiple receptor classes to many essential cellular functions, and is perhaps the most commonly activated signalling pathway in human cancer. This pathway therefore presents both
Causal Network? Modeling identifies common and unique mechanisms for sensitivity to the PI3K inhibitor GSK1059615 and the AKT inhibitor GSK690693
Macoritto M P
Molecular Cancer Therapeutics, 8(12 Suppl), 15-19 (2009)
Steven D Knight et al.
ACS medicinal chemistry letters, 1(1), 39-43 (2010-04-08)
Phosphoinositide 3-kinase α (PI3Kα) is a critical regulator of cell growth and transformation, and its signaling pathway is the most commonly mutated pathway in human cancers. The mammalian target of rapamycin (mTOR), a class IV PI3K protein kinase, is also

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service