Skip to Content
Merck
All Photos(1)

Key Documents

SMB01055

Sigma-Aldrich

Colladin

≥90% (LC/MS-ELSD)

Synonym(s):

Coladin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32O5
CAS Number:
Molecular Weight:
424.53
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

424.53

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1c2c(ccc(c2)OC[C@H]3[C@]4([C@@H](C([C@@H](CC4)OC(=O)C)(C)C)CCC3=C)C)C=CC1=O

InChI

1S/C26H32O5/c1-16-6-10-22-25(3,4)23(30-17(2)27)12-13-26(22,5)20(16)15-29-19-9-7-18-8-11-24(28)31-21(18)14-19/h7-9,11,14,20,22-23H,1,6,10,12-13,15H2,2-5H3/t20-,22-,23-,26+/m1/s1

InChI key

MNGYOFNIAOWXIT-DNRBGDKYSA-N

General description

Colladin, a sesquiterpenoid coumarin, is a natural product commonly available from Ferula Sp. (F. arrigonii, F. vesceritensis, F. campestris and F. sinaica) plants. Existing research suggests that this plant-derived metabolite exerts various biological activities, including anticancer, neuroprotective, and antiglaucoma properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

According to the existing research, Coladin exhibited notable anti-cancer properties by substantially reducing cell proliferation and mitochondrial dehydrogenase activity in mouse B16F1 melanoma cells. These effects induced apoptosis through mechanisms involving decreased mitochondrial membrane potential and mitochondrial respiratory rate, underscoring its potential as a natural anti-cancer agent. Isolated from the roots of Ferulago campestris, coladin, along with other compounds such as umbelliprenin and epielmanticine, displayed inhibitory activity against Acetylcholinesterase (AChE) with an IC50 of 0.1 mM, suggesting its potential application in the treatment of neurological disorders like Alzheimer′s disease. Coladin, identified as one of the constituents in Heptaptera triquetra fruit, exhibited remarkable inhibitory activity against acetylcholinesterase (AChE), human carbonic anhydrase isoenzyme (hCA) I, and hCA II, with IC50 values of 8.25 nM, 28.90 nM, and 43.31 nM, respectively. This highlights its potential in the development of phytotherapeutics for conditions like Alzheimer′s disease (AD) and glaucoma.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phytochemical content and enzyme inhibitory effect of Heptaptera triquetra (Vent.) Tutin fruit against acetylcholinesterase and carbonic anhydrase I and II isoenzymes
Kaya AC, et al.
Chemical Papers, 77, 5829?5837-5829?5837 (2023)
Identification of non-alkaloid acetylcholinesterase inhibitors from Ferulago campestris (Besser) Grecescu (Apiaceae)
Dall'Acqua S, et al.
Fitoterapia, 81, 1208-1212 (2010)
Cytotoxicity of sesquiterpenes ferulenol and coladin on liver FAO and B16F1 melanoma cells
Boulmeltout M, et al.
Pharmacognosy magazine, 14, 333-337 (2018)
Sesquiterpene esters and sesquiterpene coumarin ethers from Ferula linkii-TF
Gonzalez A G, et al.
Phytochemistry, 33(4), 863-866 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service