Skip to Content
Merck
All Photos(1)

Key Documents

SMB00091

Sigma-Aldrich

Altholactone

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O4
CAS Number:
Molecular Weight:
232.23
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]2OC(=O)C=C[C@@H]2O[C@@H]1c3ccccc3

InChI

1S/C13H12O4/c14-10-7-6-9-13(17-10)11(15)12(16-9)8-4-2-1-3-5-8/h1-7,9,11-13,15H/t9-,11+,12+,13+/m0/s1

InChI key

ZKIRVBNLJKGIEM-WKSBVSIWSA-N

General description

Natural product derived from plant source.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William P Unsworth et al.
Chemical communications (Cambridge, England), 47(27), 7659-7661 (2011-06-15)
We describe a concise synthesis of (+)-isoaltholactone via a Au-catalysed cyclisation of a monoallylic diol to form the tetrahydrofuranyl ring. Analogous cyclisations show that the stereochemical outcome is dictated by the stereochemistry of the diol substrate.
Shuchi Gupta et al.
Bioorganic & medicinal chemistry, 18(2), 849-854 (2009-12-29)
A related series of styryllactones with small functional and stereochemical variations were compiled for a comparative study of their apoptotic activities toward two tumorigenic and one non-tumorigenic control cell line. While a substantial range of intrinsic activity was observed, the
Dieter Enders et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(9), 2842-2849 (2008-01-26)
The asymmetric total synthesis of (+)-altholactone (1), a member of the styryllactone family of natural products displaying cytotoxic and antitumor activities, is described. Key steps include a RAMP-hydrazone alpha-alkylation (RAMP=(R)-1-amino-2-methoxymethylpyrrolidine) of 2,2-dimethyl-1,3-dioxan-5-one, a boron-mediated aldol reaction, a six- to five-membered
Kavirayani R Prasad et al.
The Journal of organic chemistry, 73(1), 2-11 (2007-05-26)
Stereoselective total synthesis of biologically active styryllactones 7-epi-goniofufurone, goniofufurone, goniopypyrone, goniotriol, altholactone, and etharvensin was achieved in high overall yields from a common intermediate derived from d-(-)-tartaric acid. It is based on the utility of a masked tetrol, comprising an
Nizar M Mhaidat et al.
Phytotherapy research : PTR, 26(6), 926-931 (2011-11-23)
Resistance of colorectal cancer (CRC) to the available chemotherapy reveals the demand for identification of new anticancer agents. We evaluated the antitumour potential of altholactone, a naturally occurring bioactive compound isolated from Goniothalamus spp. (Annonaceae) hooks, against CRC cells. Antitumour

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service