Skip to Content
Merck
All Photos(1)

Documents

SMB00079

Sigma-Aldrich

Ricinine

≥95% (LC/MS-ELSD)

Synonym(s):

Recinine, Ricidine, Ricinin, Ritsinin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2O2
CAS Number:
Molecular Weight:
164.16
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COC1=C(C#N)C(=O)N(C)C=C1

InChI

1S/C8H8N2O2/c1-10-4-3-7(12-2)6(5-9)8(10)11/h3-4H,1-2H3

InChI key

PETSAYFQSGAEQY-UHFFFAOYSA-N

General description

Ricinine is an alkaloidal toxin primarily sourced from castor beans (Ricinus communis). It belongs to the piperidine alkaloids group and contains a cyano group in its structure that contributes to its toxicity.

Biochem/physiol Actions

Dueto its toxicity, ricinine possesses insecticidal, antibacterial, and antifungalproperties against many species. It serves as a preliminary biomarker for therapid detection of ricin in blood and urine samples.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Wen et al.
Journal of proteomics, 201, 27-36 (2019-04-08)
Low temperatures profoundly influence the physiological and behavioural processes of ectotherms, especially teleosts, which have made them the subjects of strong interest over time. However, the characteristics of fish cold-tolerance at the protein level remain unclear. Therefore, to shed further
Munmi Majumder et al.
Scientific reports, 9(1), 14493-14493 (2019-10-12)
Medicinal plant-based therapies can be important for treatment of cancer owing to high efficiency, low cost and minimal side effects. Here, we report the anti-cancer efficacy of Ricinus communis L. fruit extract (RCFE) using estrogen positive MCF-7 and highly aggressive
Nick Verougstraete et al.
Journal of analytical toxicology, 43(3), e1-e5 (2018-12-28)
Ricin is a highly toxic agent derived from the castor bean plant (Ricinus communis). Poisoning occurs commonly by oral ingestion of the beans. Injection of ricin is believed to be more lethal. Ricin is a large glycosylated protein difficult to
Chang-Cai Liu et al.
Journal of proteome research, 20(1), 369-380 (2020-10-28)
Ricin is a type II ribosome-inactivating protein toxin consisting of A and B chains linked by one interchain disulfide bond. Because of its high toxicity depending on both chains together, confirming the presence of both A and B chains of
Long-Hui Liang et al.
Analytical and bioanalytical chemistry, 413(2), 585-597 (2020-11-14)
The toxic protein of ricin has drawn wide attention in recent years as a potential bioterrorism agent due to its high toxicity and wide availability. For the verification of the potential anti-terrorism activities, it is urgent for the quantification of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service