Skip to Content
Merck

SHPH15

MISSION® LentiPlex® Human Pooled shRNA Library TRC1.5

For Rapid, Convenient Genome-wide shRNA Screens

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
41105904
Concentration:
≥5x108 VP/ml (via p24 assay)
Technique(s):
capture ELISA: 5 x 108 TU/mL using p24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

MISSION®

concentration

≥5x108 VP/ml (via p24 assay)

technique(s)

capture ELISA: 5 x 108 TU/mL using p24

shipped in

dry ice

storage temp.

−70°C

General description

MISSION LentiPlex Pooled Libraries bring the power of the MISSION TRC shRNA collection together with Sigma′s lentiviral manufacturing expertise to enable genome-wide RNAi screens at your bench-top. The LentiPlex pooled system provides enhanced delivery and long-term gene silencing in non-dividing and primary cell lines allowing unlimited discovery potential. The convenient format now makes rapid whole-genome RNAi screens accessible to any researcher with minimal reagent, time or capital equipment investment.

The MISSION LentiPlex Human shRNA Pooled Library 1.5 includes additional gene and clone coverage from the previously offered LentiPlex 1.0 Pooled Library . With the addition of 1.5, which consists of 4,000+ human genes and 10,000+ total clone coverage divided into 3 subpools, the complete whole-genome human library complete whole-genome human librarynow consists of over 110,000 shRNA constructs from the TRC collection targeting 21,000+ human genes. Each library is tested for shRNA representation before product release to ensure robust library coverage. The complete library is provided in ready-to-use lentiviral format at titers of at least 5 x 108 TU/ml via p24 assay and is pre-divided into sixteen subpools of approximately 6,000-8,000 shRNA constructs each. Amplification and sequencing primers are also provided for downstream target identification.

For more information including sample data and uses, please visit our LentiPlex Technical Detail Page.

Other Notes

Browse additional MISSION shRNA Gene Family Sets in our MISSION shRNA portal.
The MISSION LentiPlex Human Pooled shRNA Libraries consist of Catalog Numbers SHPH01, SHPH15, SHPH2, and SHPHLIBR. Please refer to Table 1 for approximate number of shRNA clone coverage, targeted genes, and subpools for each library.

Amplification and sequencing primers are also provided for downstream hit identification.

Deconvolution of shRNA Pools
Sigma′s deconvolution service lets you easily identify the genes that impact your pooled shRNA screen.

  • Next-generation sequencing of clonesgives a precise number of individual clone occurrence within a pooled shRNA sample
  • Comprehensive, reproducible results from pooled shRNA screens
  • Statistically robust and information-rich data

Contact your local Sigma sales representative for more information or submit an inquiry to MISSION RNAi

Legal Information

LentiPlex is a registered trademark of Merck KGaA, Darmstadt, Germany
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library