Skip to Content
Merck

SCP0133

Endomorphin II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H37N5O5
Molecular Weight:
571.67
NACRES:
NA.32
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Application

Endomorphin-1 (Tyr-Pro-Trp-Phe-NH2, YPWF-NH2) and endomorphin-2 (Tyr-Pro-Phe-Phe-NH2, YPFF-NH2) are endogeneous opioid tetrapeptides with high affinity and specificity for the μ (mu) opioid receptor. Endomorphin-1 and 2 may be used to study the differential action and metabolism of these opioids.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library