Skip to Content

SAE0171

Angiotensin Converting Enzyme-2, ACE2

biotin-tagged, human recombinant, ≥1000000U/mg, expressed in HEK 293 cells

Synonym(s):

ACE-related carboxypeptidase, ACE2, Angiotensin-converting enzyme homolog (ACEH), COVID-19 receptor, Metalloprotease MPROT15, SARS-COV2 receptor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.83
UNSPSC Code:
12352204
Assay:
≥95% (SDS-PAGE)
Recombinant:
expressed in HEK 293 cells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in HEK 293 cells

Quality Segment

description

Biotinylation: ≥90% (gel shift assay)

assay

≥95% (SDS-PAGE)

specific activity

≥1000000 U/mg

shipped in

wet ice

storage temp.

−20°C

General description

Angiotensin Converting Enzyme-2 (ACE2) is a membrane-associated and secreted enzyme that is principally expressed on endothelium. In humans, ACE2 occurs mainly in endothelium of heart, kidney, and testis, with other expression in coronary vessel smooth muscle and kidney tubular epithelium ACE2 is a component of the renin-angiotensin system (RAS) that provides protective effects in peripheral tissues.
Coronaviruses such as SARS-CoV-2 and SARS-CoV-1 use the host ACE2 as a co-receptor to gain intracellular entry into the lungs and brain. The virion expresses a protein termed spike, which directly binds to the extracellular domain of ACE2. A specific region in the spike protein serves as the receptor binding domain (RBD). The ACE2:spike interaction has a high affinity of 15 nM.

Recombinant human ACE2 is expressed in human HEK 293 cells as a glycoprotein with a C-terminal FLAG® tag and his-tag with a calculated molecular mass of 85.9 kDa. The DTT-reduced protein migrates as a 90–120 kDa polypeptide on SDS-PAGE due to glycosylation.

This ACE2 is labelled with biotin, to form Angiotensin Converting Enzyme-2. This ACE2 biotin-tagged may serve as a useful tool for binding experiments while detecting protein: protein interaction assays using binding partners such as CoV spike protein. The degree of biotinylation is ≥90%. This ACE2-biotin can be visualized with streptavidin-conjugated probes (e.g., Streptavidin, HRP conjugate catalog No. 18-152).

Application

  • Exploration of the protein protein interaction of ACE2 with is partners, like COV Spike protein.
  • Tool for screening inhibitors for the interaction of ACE2 with COV Spike protein

Biochem/physiol Actions

Angiotensin Converting Enzyme-2 (ACE2) is a membrane-associated and secreted enzyme, that is expressed predominantly on endothelium. In humans, it is highly restricted to heart, kidney, and testis.
ACE2 is a new component of the renin-angiotensin system (RAS). It provides protective effects in peripheral tissues and has great potential for the treatment of RAS-related diseases.
Coronaviruses such as SARS-CoV-2 and SARS-CoV-1 use the host ACE2 as a coreceptor to gain intracellular entry into the lungs and brain. The virion expresses a protein termed spike, which directly binds to the extracellular domain of ACE2. A specific region in the spike protein serves as the receptor binding domain (RBD). The ACE2:spike interaction has a high affinity of 15 nM.

Features and Benefits

Our biotin-tagged ACE2 enables exploration of the ACE2 interaction with Spike protein of SARS-COV-2

Other Notes

One unit is defined as the amount of enzyme required to cleave 1 picomole of the fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH in one minute, in 37 °C, pH 7.5.
This product is supplied as a powder, lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4. The specific activity of recombinant ACE2 is measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library