Skip to Content
Merck
All Photos(2)

Key Documents

SAE0077

Sigma-Aldrich

MMP-9 human

recombinant, ≥1,300 pmol/min/μg, expressed in HEK 293 cells

Synonym(s):

GELBCLG4B, Gelatinase, Gelatinase B, MANDP2, MMP-9, Matrix Metalloproteinase-9, Type IV collagenase

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

95% (SDS-PAGE)

form

liquid

mol wt

calculated mol wt 76 kDa
observed mol wt 92 kDa (The protein migrates as a 92 kDa protein on SDS-PAGE due to glycosylation)

concentration

50-200 μg/mL

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MMP(4318)

General description

50 μg protein, determined by Bradford. For the lot-specific concentration, see Certificate of Analysis.
Recombinant human Matrix Metalloproteinase-9 (MMP-9) is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of 76 kDa (amino acids 20-707). The DTT-reduced protein migrates as a ~92 kDa polypeptide on SDS-PAGE due to glycosylation. This protein is manufactured in human cells, with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports higher specific activity of the protein. The protein is produced with no artificial tags.

Biochem/physiol Actions

MMP-9 is a member of the matrix metalloproteinase (MMP) family of proteins. Proteins of the MMP family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Studies in rhesus monkeys suggest that MMP9 is involved in IL-8 (interleukin-8)-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling. Thrombospondins, intervertebral disc proteins, regulate the effective levels of MMP-2 and -9, which are key effectors of extracellular matrix (ECM) remodeling. MMP-9 degrades various substrates including gelatin, collagen types IV and V, and elastin. MMP-9 is involved in a variety of autoimmune diseases such as systemic lupus erythematosus, rheumatoid arthritis, and multiple sclerosis, and be regarded as a potential therapeutic target.
MMP-9 is secreted from neutrophils, macrophages, and a number of transformed cells, and is the most complex family member in terms of domain structure and regulation of its activity.
Structurally, MMP9 maybe be divided into five distinct domains: a pro-domain which is cleaved upon activation, a gelatin binding domain consisting of three contiguous fibronectin type II units, a catalytic domain containing the zinc binding site, a proline rich linker region, and a carboxyl terminal hemopexin like domain.

Other Notes

This product can be activated in vitro by adding 4-Aminophenylmercuric acetate (APMA), Cat. No. A9563, to a final concentration of 1 mM.

Physical form

Liquid solution, 0.22 mm filtered, containing 25 mM Tris, 10 mM CaCl2, 150 mM NaCl, 0.05% Brij-35

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamara Filipović et al.
BioMed research international, 2020, 9758289-9758289 (2020-02-20)
Osteoporosis is a disease characterized by decreased bone density and destruction of bone microarchitecture. Indicators for altered bone homeostasis are changes in the serum level of matrix metalloproteinases (MMPs) and their tissue inhibitors (TIMPs). The purpose of the current study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service