SAB5900404
Anti-Monkeypox virus/MPXV E8L Antibody (SAA0285)
Synonym(s):
E8L, Cell surface-binding protein, Carbonic anhydrase homolog, Monkeypox virus (MPXV)
About This Item
Recommended Products
biological source
mouse
Quality Level
recombinant
expressed in CHO cells
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
clone
SAA0285, recombinant monoclonal
Assay
>95% (SDS-PAGE)
form
liquid
mol wt
150 kDa
species reactivity
monkeypox virus
concentration
1 mg/mL
technique(s)
ELISA: 1:2000-1:4000
neutralization: 0.01-10 μg/mL
color
colorless
pH
7.4
isotype
IgG2a, kappa
Protein ID accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
-65 to -96°C
target post-translational modification
unmodified
General description
Monkeypox virus (MPXV), like smallpox virus, belongs to the genus Orthopoxvirus. Its morphology is consistent with that of orthopoxvirus, with a brick-shaped or oval shape with rounded corners and a size of 200~400nm. Brick-shaped or oval-shaped, covered by a lipoprotein membrane or envelope, with two protein-containing side bodies in the middle, and a thick membrane core containing a large double-stranded DNA gene body. The genome of Mpox virus is a double-stranded DNA, about 197kb long, and the end of the genome contains an identical but opposite terminal inverted repeat sequence. The virus contains 190 open reading frames, 4 of which are located in terminal inverted repeats. The core proteins of monkeypox virus are A29L, A35R, B6R, M1R, H3L.
Specificity
Application
Packaging
Storage and Stability
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.
Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.
Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.
Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service