SAB5500139
Anti-MAGE-A1 antibody, Rabbit monoclonal
recombinant, expressed in proprietary host, clone SP188, affinity isolated antibody
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in proprietary host
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
SP188, monoclonal
species reactivity
human (tested)
technique(s)
flow cytometry: 1:400
immunoblotting: 1:400
immunohistochemistry: 1:100
isotype
IgG
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... MAGEA1(4100)
Related Categories
General description
Melanoma-associated antigen 1 (MAGE-A1) is one of the melanoma associated antigens (MAAs). MAGE-A1 is highly expressed in testis and some human solid tumors such as melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian and renal cell carcinomas.
Immunogen
Synthetic peptide derived from the C-terminus of human MAGE-A1 protein.
Biochem/physiol Actions
Members of MAGE family play a vital role in maintaining normal cellular physiology. Melanoma-associated antigen 1 (MAGE-A1) serves as a target antigen of patient-derived CD8+ T cells in melanoma. It might be potential target for treating melanoma patients.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
MAGE-F1, a novel ubiquitously expressed member of the MAGE superfamily.
Stone B, et al.
Gene, 267(2), 173-182 (2001)
MAGE-A1 promotes melanoma proliferation and migration through C-JUN activation.
Wang D, et al.
Biochemical and Biophysical Research Communications, 473(4), 959-965 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service