Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200093

Sigma-Aldrich

Anti-Methylated Lysine antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Methylated Lysine, Anti-N epsilon dimethyl lysine, Anti-methyl lysine, Anti-trimethyl lysine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

several mammalian species

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

methylation (Lys)

General description

Methylated lysine is a methyl derivative of lysine and is present in muscle proteins.

Specificity

Detects proteins containing methylated lysine residues in SDS PAGE immunoblots.

Immunogen

Methylated KLH Conjugated

Application

Anti-Methylated Lysine antibody produced in rabbit has been used in western blotting.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, 50% glycerol, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amino Acid Modifications
Engelking, Larry R.
Textbook of Veterinary Physiological Chemistry, 12-17 (2015)
Rodney F Minchin et al.
Biochemical pharmacology, 158, 153-160 (2018-10-21)
In the present study, a screen of adenosine analogs as potential modulators of arylamine-N-acetyltransferase 1 activity identified ATP as an inhibitor within its range of physiological concentrations. Kinetically, ATP was a non-competitive inhibitor with respect to the acetyl acceptor but
Maria E Gonzalez et al.
iScience, 25(8), 104827-104827 (2022-08-23)
Triple-negative breast cancers (TNBCs) are frequently poorly differentiated with high propensity for metastasis. Enhancer of zeste homolog 2 (EZH2) is the lysine methyltransferase of polycomb repressive complex 2 that mediates transcriptional repression in normal cells and in cancer through H3K27me3.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service